Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200106

Sigma-Aldrich

Anti-ULK1 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-ATG1 autophagy related 1 homolog, Anti-UNC51, Anti-Unc51.1, Anti-unc-51-like kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of HEK-293T cells overexpressing human ULK1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULK1(8408)

General description

Uncoordinated 51-like kinase 1 (ULK1) is expressed in skeletal muscle, heart, pancreas, brain, placenta, liver, kidney and lung. This gene is located on human chromosome 2q24.3. ULK1 localizes to cytoplasmic structures. ULK1 and ULK2 is a homolog of autophagy related 1 (Atg1) gene.

Specificity

Anti-ULK1 recognizes human ULK1.

Application

Anti-ULK1 has been used in immunoblotting.

Biochem/physiol Actions

Uncoordinated 51-like kinase 1 (ULK1) is involved in modulating Atg9 subcellular dynamics. Knockdown of ULK1, but not ULK2, inhibits the autophagic response. Overexpression of ULK1 also inhibits autophagy, suggesting that ULK1 may have multiple mechanisms for regulating autophagy. Knockdown of Ulk1 shRNA can prevent gastric cancer cell survival.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemcitabine induces poly (ADP-ribose) polymerase-1 (PARP-1) degradation through autophagy in pancreatic cancer
Wang Y, et al.
PLoS ONE, 9(10), e109076-e109076 (2014)
siRNA screening of the kinome identifies ULK1 as a multidomain modulator of autophagy
Chan EYW, et al.
The Journal of Biological Chemistry, 282(35), 25464-25474 (2007)
Ulk1 over-expression in human gastric cancer is correlated with patients' T classification and cancer relapse
Chen MB, et al.
Testing, 8(20), 33704-33704 (2017)
Human ULK1, a Novel Serine/Threonine Kinase Related to UNC-51 Kinase ofCaenorhabditis elegans: cDNA Cloning, Expression, and Chromosomal Assignment
Kuroyanagi H, et al.
Genomics, 51(1), 76-85 (1998)
Xin Yu et al.
Journal of molecular neuroscience : MN, 50(3), 586-599 (2013-04-18)
Spinocerebellar ataxia type 7 (SCA7) is one of nine neurodegenerative disorders caused by expanded polyglutamine domains. These so-called polyglutamine (polyQ) diseases are all characterized by aggregation. Reducing the level of aggregating polyQ proteins via pharmacological activation of autophagy has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service