Skip to Content
Merck
All Photos(4)

Documents

SAB2104467

Sigma-Aldrich

Anti-ETV1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp781L0674, Anti-ER81, Anti-MGC104699, Anti-MGC120533, Anti-MGC120534

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

dog, horse, mouse, guinea pig, human, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETV1(2115)

General description

Ets variant gene 1 (ETV1) is a novel androgen-regulated gene mapped to human chromosome 7p21.2. ETV1 protein belongs to the E-twenty-six (ETS) transcription factor family and it contains the ETS domain and acidic transactivation domain. ETV1 binds to DNA sequences containing the consensus pentanucleotide 5′-CGGA[AT]-3′.

Immunogen

Synthetic peptide directed towards the middle region of human ETV1

Application

Anti-ETV1 antibody produced in rabbit has been used in western blot(1:500)

Biochem/physiol Actions

E-twenty-six (ETS) proteins participate in cell proliferation and cancer cell invasion. Ets variant gene 1 (ETV1) acts as a neuregulin-1 (NRG1) responsive factor and is important for establishing rapid conduction physiology in the heart. Overexpression of the gene has been observed in various types of cancers including prostate cancer and gastrointestinal stromal tumors

Sequence

Synthetic peptide located within the following region: PDNQRPLLKTDMERHINEEDTVPLSHFDESMAYMPEGGCCNPHPYNEGYV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ETV1 is a novel androgen receptor-regulated gene that mediates prostate cancer cell invasion.
Cai, et al.
Molecular Endocrinology, 21, 1835-1846 (2013)
Ping Chi et al.
Nature, 467(7317), 849-853 (2010-10-12)
Gastrointestinal stromal tumour (GIST) is the most common human sarcoma and is primarily defined by activating mutations in the KIT or PDGFRA receptor tyrosine kinases. KIT is highly expressed in interstitial cells of Cajal (ICCs)-the presumed cell of origin for
Mark A Rubin et al.
Laboratory investigation; a journal of technical methods and pathology, 86(11), 1099-1102 (2006-09-20)
Recurrent chromosomal rearrangements have not been well characterized in common carcinomas. We describe the use of a novel bioinformatics approach to discover candidate oncogenic chromosomal aberrations on the basis of outlier gene expression called COPA (cancer outlier profile analysis). We
Xiao Song et al.
Cancer research, 79(20), 5288-5301 (2019-08-30)
Misregulated alternative RNA splicing (AS) contributes to the tumorigenesis and progression of human cancers, including glioblastoma (GBM). Here, we showed that a major splicing factor, serine and arginine rich splicing factor 3 (SRSF3), was frequently upregulated in clinical glioma specimens
La Ta et al.
Molecular medicine reports, 14(2), 1371-1378 (2016-06-10)
Constitutive photomorphogenic 1 (COP1) belongs to the COP‑de-etiolated (DET)‑fusca (FUS) protein family and has been demonstrated to suppress prostate adenocarcinomas and other types of tumor, such as liver and gastric cancer. The present study investigated the expression of COP1 and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service