Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411231

Sigma-Aldrich

Anti-CCL2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 11 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL2(6347)

General description

C-C motif chemokine ligand 2 (CCL2) gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4. (provided by RefSeq).CCL2 is a pro-inflammatory cytokine. It belongs to the family of CC-type chemokines, which can be produced by almost every cells.

Immunogen

CCL2 (NP_002973.1, 1 a.a. ~ 99 a.a) full-length human protein.

Sequence
MKVSAALLCLLLIAATFIPQGLAQPDAINAPVTCCYNFTNRKISVQRLASYRRITSSKCPKEAVIFKTIVAKEICADPKQKWVQDSMDHLDKQTQTPKT

Biochem/physiol Actions

C-C motif chemokine ligand 2 (CCL2) plays a critical role in the development of allergic inflammatory reactions by employing immune cells, that is linked to autoimmune diseases. It binds to CC chemokine receptor 2 (CCR2) and regulates monocyte chemotaxis and T-lymphocyte differentiation. The protein recruits monocytes to liver. It is involved in macrophage fusion and foreign body giant cell formation.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The regulation and importance of monocyte chemoattractant protein-1
Bianconi V, et al.
Current Opinion in Hematology, 25(1), 44-51 (2018)
Association of MCP-1 rs1024611 polymorphism with diabetic foot ulcers
Su N, et al.
Medicine, 97(28) (2018)
Monocyte chemoattractant protein 1 released from macrophages induced by hepatitis C virus promotes monocytes migration
Liu Y, et al.
Virus Research, 240, 190-196 (2017)
The CC chemokine ligand, CCL2/MCP1, participates in macrophage fusion and foreign body giant cell formation
Kyriakides TR, et al.
The American Journal of Pathology, 165(6), 2157-2166 (2004)
Monocyte chemoattractant protein-1 (MCP-1)-2518 A/G polymorphism and lupus nephritis risk: A PRISMA-compliant meta-analysis
Sang GY, et al.
Medicine, 96(51) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service