Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409921

Sigma-Aldrich

Anti-LY6D antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

E48

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 13.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LY6D(8581)

General description

The gene LY6D (lymphocyte antigen 6 complex, locus D) is mapped to human chromosome 8q24.3. It belongs to the LY6 (lymphocyte antigen 6) gene family. The encoded protein is a membrane protein with molecular weight of 14kDa. It is a glycosyl phosphatidylinositol (GPI)-anchored protein.

Immunogen

LY6D (NP_003686.1, 1 a.a. ~ 128 a.a) full-length human protein.

Sequence
MRTALLLLAALAVATGPALTLRCHVCTSSSNCKHSVVCPASSRFCKTTNTVEPLRGNLVKKDCAESCTPSYTLQGQVSSGTSSTQCCQEDLCNEKLHNAAPTRTALAHSALSLGLALSLLAVILAPSL

Biochem/physiol Actions

LY6D (lymphocyte antigen 6 complex, locus D) is required for discriminating B lineage restricted common lymphoid progenitors. It is upregulated by X-ray irradiation-mediated DNA damage pathway controlled via ATM (ataxia telangiectasia mutated), CHK2 (checkpoint kinase 2) and p53. LY6D is upregulated in various cancers and is associated with patient survival outcome.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingzhao Zhang et al.
PloS one, 8(8), e72397-e72397 (2013-09-12)
Common lymphoid progenitors (CLPs) are thought to represent major intermediates in the transition of hematopoietic stem cells (HSCs) to B lineage lymphocytes. However, it has been obvious for some time that CLPs are heterogeneous, and there has been controversy concerning
Jie Hu et al.
American journal of medical genetics. Part A, 167A(8), 1921-1926 (2015-04-14)
A 7-year-old female with developmental delay (DD), autism spectrum disorder (ASD), intellectual disability (ID), attention deficit hyperactivity disorder (ADHD), and seizures was referred to our laboratory for oligomicroarray analysis. The analysis revealed a 540 kb microdeletion in the chromosome 8q24.3 region
Maiko Kurosawa et al.
The FEBS journal, 279(24), 4479-4491 (2012-10-19)
In order to identify membrane proteins whose expression is induced by X-ray irradiation, we developed an antibody (Ab)-directed strategy using a phage Ab library. X-Ray-irradiated cells were screened with a phage Ab library in the presence of a large excess
Linlin Luo et al.
Oncotarget, 7(10), 11165-11193 (2016-02-11)
Stem cell antigen-1 (Sca-1) is used to isolate and characterize tumor initiating cell populations from tumors of various murine models [1]. Sca-1 induced disruption of TGF-β signaling is required in vivo tumorigenesis in breast cancer models [2, 3-5]. The role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service