Skip to Content
Merck
All Photos(2)

Key Documents

SAB1407412

Sigma-Aldrich

Anti-SMARCAL1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HARP, HHARP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~105.9 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein shows sequence similarity to the E. coli RNA polymerase-binding protein HepA. Mutations in this gene are a cause of Schimke immunoosseous dysplasia (SIOD), an autosomal recessive disorder with the diagnostic features of spondyloepiphyseal dysplasia, renal dysfunction, and T-cell immunodeficiency. (provided by RefSeq)

Immunogen

SMARCAL1 (NP_054859.2, 1 a.a. ~ 954 a.a) full-length human protein.

Sequence
MSLPLTEEQRKKIEENRQKALARRAEKLLAEQHQRTSSGTSIAGNPFQAKQGPSQNFPRESCKPVSHGVIFKQQNLSSSSNADQRPHDSHSFQAKGIWKKPEEMPTACPGHSPRSQMALTGISPPLAQSPPEVPKQQLLSYELGQGHAQASPEIRFTPFANPTHKPLAKPKSSQETPAHSSGQPPRDAKLEAKTAKASPSGQNISYIHSSSESVTPRTEGRLQQKSGSSVQKGVNSQKGKCVRNGDRFQVLIGYNAELIAVFKTLPSKNYDPDTKTWNFSMNDYSALMKAAQSLPTVNLQPLEWAYGSSESPSTSSEGQAGLPSAPSLSFVKGRCMLISRAYFEADISYSQDLIALFKQMDSRRYDVKTRKWSFLLEEHSKLIAKVRCLPQVQLDPLPTTLTLAFASQLKKTSLSLTPDVPEADLSEVDPKLVSNLMPFQRAGVNFAIAKGGRLLLADDMGLGKTIQAICIAAFYRKEWPLLVVVPSSVRFTWEQAFLRWLPSLSPDCINVVVTGKDRLTAGLINIVSFDLLSKLEKQLKTPFKVVIIDESHFLKNSRTARCRAAMPVLKVAKRVILLSGTPAMSRPAELYTQIIAVKPTFFPQFHAFGLRYCDAKRMPWGWDYSGSSNLGELKLLLEEAVMLRRLKSDVLSQLPAKQRKIVVIAPGRINARTRAALDAAAKEMTTKDKTKQQQKDALILFFNRTAEAKIPSVIEYILDLLESGREKFLVFAHHKVVLDAITQELERKHVQHIRIDGSTSSAEREDLCQQFQLSERHAVAVLSITAANMGLTFSSADLVVFAELFWNPGVLIQAEDRVHRIGQTSSVGIHYLVAKGTADDYLWPLIQEKIKVLAEAGLSETNFSEMTESTDYLYKDPKQQKIYDLFQKSFEKEGSDMELLEAAESFDPGSASGTSGSSSQNMGDTLDESSLTASPQKKRRFEFFDNWDSFTSPL

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service