Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401383

Sigma-Aldrich

Anti-TFAM antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MtTF1, TCF6, TCF6L1, TCF6L2, TCF6L3, mtTFA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFAM(7019)

Related Categories

General description

Mitochondrial transcription factor A (TFAM) is a high-mobility group (HMG) protein and is made up of two HMG-box domains. The TFAM gene is located on the human chromosome at 10q21.1.

Immunogen

TFAM (NP_003192.1, 1 a.a. ~ 246 a.a) full-length human protein.

Sequence
MAFLRSMWGVLSALGRSGAELCTGCGSRLRSPFSFVYLPRWFSSVLASCPKKPVSSYLRFSKEQLPIFKAQNPDAKTTELIRRIAQRWRELPDSKKKIYQDAYRAEWQVYKEEISRFKEQLTPSQIMSLEKEIMDKHLKRKAMTKKKELTLLGKPKRPRSAYNVYVAERFQEAKGDSPQEKLKTVKENWKNLSDSEKELYIQHAKEDETRYHNEMKSWEEQMIEVGRKDLLRRTIKKQRKYGAEEC

Application

Anti-TFAM antibody produced in rabbit has been used in western blotting (1:1000) and immunofluorescence.

Biochem/physiol Actions

Mitochondrial transcription factor A (TFAM) is involved in mitochondrial DNA (mtDNA) synthesis, expression, and packaging. It is also involved in regulating the aggregation of mtDNA. TFAM stabilizes the mtDNA by binding to it in a sequence-depending manner and forms a nucleoid.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Alvarez et al.
Journal of Alzheimer's disease : JAD, 13(3), 275-280 (2008-04-24)
Impaired mitochondrial function and an increased number of mutations in mitochondrial DNA (mtDNA) has been found in brains of patients with late-onset Alzheimer's disease (LOAD). The TFAM-gene encodes the mitochondrial transcription factor A, a protein that controls the transcription, replication
Ryan Neil Marshall et al.
Frontiers in physiology, 13, 1097988-1097988 (2023-01-24)
Background: Ageing is associated with alterations to skeletal muscle oxidative metabolism that may be influenced by physical activity status, although the mechanisms underlying these changes have not been unraveled. Similarly, the effect of resistance exercise training (RET) on skeletal muscle
Ryan N Marshall et al.
Physiological reports, 10(13), e15345-e15345 (2022-07-06)
Bed rest (BR) results in significant impairments in skeletal muscle metabolism. Mitochondrial metabolism is reportedly highly sensitive to disuse, with dysregulated fission-fusion events and impaired oxidative function previously reported. The effects of clinically relevant short-term BR (≤5 days) on mitochondrial protein
Dongchon Kang et al.
Mitochondrion, 7(1-2), 39-44 (2007-02-07)
A growing body of evidence suggests that mammalian mitochondrial DNA takes on higher structure called nucleoid or mitochromosome corresponding to that of nuclear DNA. Mitochondrial transcription factor A (TFAM), which was cloned as a transcription factor for mitochondrial DNA, has
Phuong Xuan Tran et al.
Molecular therapy oncolytics, 24, 897-908 (2022-05-17)
For advanced oral squamous cell carcinoma (OSCC), increasing sensitivity to chemotherapy is a major challenge in improving treatment outcomes, and targeting cytoprotective processes that lead to the chemotherapy resistance of cancer cells may be therapeutically promising. Tumor-suppressive microRNAs (miRNAs) can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service