Skip to Content
Merck
All Photos(1)

Key Documents

S9564

Sigma-Aldrich

Amyloid Precursor Protein α, Secreted human

>90% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal histidine tagged), buffered aqueous solution

Synonym(s):

sAPPα

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli (N-terminal histidine tagged)

Quality Level

Assay

>90% (SDS-PAGE)

form

buffered aqueous solution

mol wt

~100 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... APP(351)

General description

The APP (amyloid precursor protein) gene is mapped to human chromosome 21q21.3. It encodes a integral membrane protein. APPα is a soluble protein generated by sequential cleavage with α and γ secretase.

Application

Human amyloid precursor protein α secreted, has been used in enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Amyloid precursor protein α is an α-secretase-cleaved soluble protein that has been shown to have neuroprotective properties. It is derived from amyloid precursor protein. The protein consists of 612 amino acids. Several G protein-coupled receptors are known to activate α-secretase-dependent processing of APP. It has neuroprotective, neurogenic and neurotrophic functions. Amyloid precursor protein a also stimulates gene expression and protein expression.

Physical form

Solution 0.2 μm filtered, in phosphate buffered saline, pH 7.4.

Preparation Note

Expressed as a soluble protein and purified under non-denaturing conditions.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dementia: New Insights for the Healthcare Professional, 478-478 (2013)
S W Barger et al.
Brain research. Molecular brain research, 40(1), 116-126 (1996-08-01)
A significant fraction of the beta-amyloid precursor protein is proteolytically processed to yield large secreted forms (sAPP). These proteins have pleiotropic effects which potentially involve control of gene expression. We have investigated the influence of sAPP on the class of
Soluble ?-amyloid Precursor Protein Alpha Binds to p75 Neurotrophin Receptor to Promote Neurite Outgrowth
Noriko Hasebe
PLoS ONE (2013)
C U Pietrzik et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(4), 1770-1775 (1998-03-21)
In various species, thyrotropin (TSH) is known to stimulate both differentiation and proliferation of thyroid follicle cells. This cell type has also been shown to express members of the Alzheimer amyloid precursor (APP) protein family and to release the secretory
SnapShot: genetics of Alzheimer?s disease
Guerreiro R, et al.
Cell, 155(4), 968-968 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service