Skip to Content
Merck
All Photos(1)

Key Documents

S8326

Sigma-Aldrich

SKI 5C

≥98% (HPLC)

Synonym(s):

2,2-Dimethyl-4S-(1-oxo-2-hexadecyn-1-yl)-1,1-dimethylethyl ester-3-oxazolidinecarboxylic acid, SPHK1 Inhibitor 5C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H45NO4
CAS Number:
Molecular Weight:
435.64
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to light brown

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC#CC(=O)[C@@H]1COC(C)(C)N1C(=O)OC(C)(C)C

InChI

1S/C26H45NO4/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-23(28)22-21-30-26(5,6)27(22)24(29)31-25(2,3)4/h22H,7-18,21H2,1-6H3/t22-/m0/s1

InChI key

YGBSGZPIDCXNEH-QFIPXVFZSA-N

Application

SKI 5C was used to inhibit Sphingosine kinase 1 in SK-Hep1, HEK-293T cells.1

Biochem/physiol Actions

SKI 5C is a selective Sphingosine Kinase 1 (SPHK1) inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youmna Atieh et al.
Current biology : CB, 31(6), 1129-1140 (2021-01-06)
Extrusion is a mechanism used to eliminate unfit, excess, or dying cells from epithelial tissues. The initial events guiding which cells will be selectively extruded from the epithelium are not well understood. Here, we induced damage in a subset of
Meiyan Bao et al.
Liver international : official journal of the International Association for the Study of the Liver, 32(2), 331-338 (2011-11-22)
Sphingosine kinase 1 (SphK1), which phosphorylates sphingosine to sphingosine-1-phosphate (S1P), is overexpressed in various types of cancers, and may act as an oncogene in tumorigenesis. However, little is known about the precise role of the SphK1/S1P pathway in human liver
Sebastian Wurster et al.
Cell reports, 34(12), 108896-108896 (2021-03-25)
Severe and often fatal opportunistic fungal infections arise frequently following mucosal damage caused by trauma or cytotoxic chemotherapy. Interaction of fungal pathogens with epithelial cells that comprise mucosae is a key early event associated with invasion, and, therefore, enhancing epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service