S7446
Superoxide Dismutase from bovine erythrocytes
BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Synonym(s):
SOD, Superoxide: superoxide oxidoreductase
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
product line
BioUltra
Assay
≥97% (SDS-PAGE)
form
lyophilized powder
specific activity
≥4,500 units/mg protein
mol wt
32.5 kDa
composition
Protein, ≥95% biuret
relevant disease(s)
arthritis
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Research area: Cell signaling. Superoxide Dismutase (SOD) is found in allaerobic cells of the animals, plants, and microorganisms. It is a low molecularweight protein that exists as three different forms such as manganese SOD,copper–zinc SOD, and extracellular SOD.
Application
Superoxide Dismutase from bovine erythrocytes has been used in a study to assess activity parameters in adjuvant-induced arthritis. Superoxide Dismutase from bovine erythrocytes has also been used in a study to simplify the process of purifying superoxide dismutases using charge-controlled hydrophobic chromatography on 10-carboxydecyl Sepharose. It has also been used as a standard in SOD assay in mice liver andkidney samples.
Biochem/physiol Actions
Superoxide Dismutase from bovine erythrocytes catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. It serves as an antioxidant and plays a critical role in the defense of cells against the toxic effects of oxygen radicals. Competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.
Unit Definition
One unit will inhibit reduction of cytochrome c by 50% in a coupled system with xanthine oxidase at pH 7.8 at 25 °C in a 3.0 mL reaction volume. Xanthine oxidase concentration should produce an initial ΔA550 of 0.025 ± 0.005 per min.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A García-González et al.
Archives of medical research, 30(2), 132-137 (1999-06-18)
The purpose of this work was to evaluate the effect of superoxide dismutase (SOD) on primary swelling, lipoperoxidation, body thymus, and spleen weight in the adjuvant-induced arthritis (AIA) model in rats. Orally and intraperitoneally administered SOD (100 U/kg) from bovine
Aloysia citrodora extract as a chemopreventive agent against HPV16-induced lesions: findings from K14-HPV16 mice
Medeiros F, et al.
Exploration of targeted anti-tumor therapy, 5, 416-433 (2024)
Purification of pro- and eukaryotic superoxide dismutases by charge-controlled hydrophobic chromatography
Grunow, M. and W. Schopp
Journal of Chromatography A, 590, 247-253 (1992)
Lu Miao et al.
Free radical biology & medicine, 47(4), 344-356 (2009-05-30)
Numerous short-lived and highly reactive oxygen species (ROS) such as superoxide (O2(.-)), hydroxyl radical, and hydrogen peroxide are continuously generated in vivo. Depending upon concentration, location, and intracellular conditions, ROS can cause toxicity or act as signaling molecules. The cellular
Karen E Butler et al.
Frontiers in chemistry, 9, 614595-614595 (2021-02-27)
Native mass spectrometry has emerged as a powerful tool for structural biology as it enables the evaluation of molecules as they occur in their physiological conditions. Ion mobility spectrometry-mass spectrometry (IMS-MS) has shown essential in these analyses as it allows
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service