S7326
SB 657510
≥98% (HPLC)
Synonym(s):
2-Bromo-N-[4-chloro-3-[[(3R)-1-methyl-3-pyrrolidinyl]oxy]phenyl]-4,5-dimethoxy-benzenesulfonamide, SB-657510
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to off-white
solubility
DMSO: ≥30 mg/mL
originator
GlaxoSmithKline
storage temp.
room temp
SMILES string
COc1cc(Br)c(cc1OC)S(=O)(=O)Nc2ccc(Cl)c(O[C@@H]3CCN(C)C3)c2
InChI
1S/C19H22BrClN2O5S/c1-23-7-6-13(11-23)28-16-8-12(4-5-15(16)21)22-29(24,25)19-10-18(27-3)17(26-2)9-14(19)20/h4-5,8-10,13,22H,6-7,11H2,1-3H3/t13-/m1/s1
InChI key
KQCZCINJGIRLCD-CYBMUJFWSA-N
Related Categories
Application
Biochem/physiol Actions
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service