Skip to Content
Merck
All Photos(1)

Key Documents

S6826

Sigma-Aldrich

Stiripentol

≥98% (HPLC)

Synonym(s):

4,4-dimethyl-1-[3,4(methylenedioxy)-phenyl]-1-penten-3-ol, Diacomit

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O3
CAS Number:
Molecular Weight:
234.29
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥20 mg/mL

storage temp.

−20°C

SMILES string

CC(C)(C)C(O)\C=C\c1ccc2OCOc2c1

InChI

1S/C14H18O3/c1-14(2,3)13(15)7-5-10-4-6-11-12(8-10)17-9-16-11/h4-8,13,15H,9H2,1-3H3/b7-5+

InChI key

IBLNKMRFIPWSOY-FNORWQNLSA-N

Related Categories

Application

Stiripentol has been used in pharmacokinetic analysis[1] and as a reference standard in fluorescence spectra analysis.[2]
Stiripentol may be used in GABAA receptor-mediated cell signaling studies.

Biochem/physiol Actions

Anti-convulsant drug

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Chiron et al.
Lancet (London, England), 356(9242), 1638-1642 (2000-11-23)
Stiripentol is an inhibitor of cytochrome P450 that showed antiepileptic efficacy in severe myoclonic epilepsy in infancy (SMEI) in association with clobazam and valproate in an open study. To confirm these results, 41 children with SMEI were included in a
Screening of conventional anticonvulsants in a genetic mouse model of epilepsy.
Hawkins NA, et al.
Annals of Clinical and Translational Neurology, 4(5), 326-339 (2017)
Carole Giraud et al.
Drug metabolism and disposition: the biological fate of chemicals, 34(4), 608-611 (2006-01-18)
A metabolic interaction between stiripentol (STP), an anticonvulsant agent that inhibits the activity of several cytochromes P450 (P450s), and clobazam (CLB), a 1,5-benzodiazepine, used in association with STP in severe myoclonic epilepsy in infancy was observed in vivo. This interaction
M Bebin et al.
Drugs, 48(2), 153-171 (1994-08-01)
In the past decade, several new antiepileptic drugs have been tested. Most recently, 5 new antiepileptic drugs have been launched onto European and US markets. These include vigabatrin, oxcarbazepine and lamotrigine in Europe, and felbamate and gabapentin in the US.
Behrouz Kassaï et al.
Epilepsia, 49(2), 343-348 (2007-11-22)
Severe myoclonic epilepsy in infancy (SMEI) is a rare, but severe disorder with seizures typically resistant to conventional antiepileptic drugs. The objective of the present study was to systematically review the literature on the available treatments for SMEI. Databases searched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service