Skip to Content
Merck
All Photos(1)

Key Documents

S5933

Sigma-Aldrich

Anti-STAT3 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 92 kDa

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: 10 μg/mL using A431 cells
immunoprecipitation (IP): 4 μg using STAT3 from 0.5–1 mg of an EGF stimulated human A431 cell lysate
western blot (chemiluminescent): 0.5-2 μg/mL using lysates from EGF stimulated human A431, mouse WEHI and rat L6 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)
mouse ... Stat3(20848)
rat ... Stat3(25125)

General description

Signal transducer and activator of transcription 3 (STAT3) protein is a member of the STAT family of proteins. It consists of a DNA binding domain, a Src homology-2 (SH-2) domain, an N-terminal coiled-coiled domain, and a C-terminal transactivation domain. The STAT3 gene is located on the human chromosome at 17q21.2.

Specificity

The antibody is specific for STAT3. It may detect an additional 66 kDa band using high concentrations or long exposures. The sequence recognized by the antibody (688-722) is identical in mouse and rat STAT3. The first 28 amino acids are identical to mouse STAT3B.

Immunogen

GST fusion protein corresponding to amino acids 688-722 of human STAT3, expressed in bacteria.

Application

Anti-STAT3 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Signal transducer and activator of transcription 3 (STAT3) signaling participates in cancer inflammation. This protein interacts with nuclear factor-κB (NF-κB). STAT3 regulates the function of several genes that take part in proliferation, apoptosis, metastasis, angiogenesis, immune response, and cell differentiation. Overexpression of the STAT3 gene is associated with lymphomas, prostate carcinoma, breast cancer, hepatocellular carcinoma, pancreatic adenocarcinoma, cholangiocarcinoma, and ovarian carcinoma.

Physical form

Solution in 0.1 M Tris-glycine, pH 7.4, containing 0.15 M NaCl and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Jin et al.
Stem cell research, 41, 101586-101586 (2019-11-11)
Autosomal dominant Hyper IgE syndrome (AD-HIES), a rare immune deficiency affecting fewer than one per million people, is caused by heterozygous deleterious mutations in STAT3. STAT3 signaling plays crucial roles in basic cellular functions affecting broad aspects of cellular homeostasis.
Hua Yu et al.
Nature reviews. Cancer, 9(11), 798-809 (2009-10-24)
Commensurate with their roles in regulating cytokine-dependent inflammation and immunity, signal transducer and activator of transcription (STAT) proteins are central in determining whether immune responses in the tumour microenvironment promote or inhibit cancer. Persistently activated STAT3 and, to some extent
Paul A Johnston et al.
Molecular interventions, 11(1), 18-26 (2011-03-29)
Multiple lines of evidence place STAT3 at a central node in the development, progression, and maintenance of many human tumors, and STAT3 has been validated as an anti-cancer target in several contexts. STAT3 modulates the transcription of a variety of
A Furuhata et al.
Biochemical and biophysical research communications, 383(4), 411-416 (2009-04-15)
Janus kinase 2 (JAK2) V617F mutation has been regarded as the major cause of myeloproliferative disorders (MPD). However, the mechanisms of abnormal cell growth by JAK2V617F have not been elucidated. In this study, cell cycle regulatory protein expression was analyzed
Mohamed Jeljeli et al.
Cell reports, 33(5), 108325-108325 (2020-11-05)
Endometriosis is a frequent, chronic, inflammatory gynecological disease characterized by the presence of ectopic endometrial tissue causing pain and infertility. Macrophages have a central role in lesion establishment and maintenance by driving chronic inflammation and tissue remodeling. Macrophages can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service