Skip to Content
Merck
All Photos(1)

Documents

S5451

Sigma-Aldrich

Sarpogrelate hydrochloride

≥98% (HPLC)

Synonym(s):

Anplag, Butanedioic acid 1-[2-(dimethylamino)-1-[[2-[2-(3-methoxyphenyl)ethyl]phenoxy]methyl]ethyl] ester hydrochloride, MCI 9042, Succinic acid mono-(1-dimethylaminomethyl-2-(2-[2-(3-methoxyphenyl)et hyl]phenoxy)ethyl) ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31NO6.HCl
CAS Number:
Molecular Weight:
465.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

deionized water: ≥5 mg/mL

storage temp.

room temp

SMILES string

Cl.COc1cccc(CCc2ccccc2OCC(CN(C)C)OC(=O)CCC(O)=O)c1

InChI

1S/C24H31NO6.ClH/c1-25(2)16-21(31-24(28)14-13-23(26)27)17-30-22-10-5-4-8-19(22)12-11-18-7-6-9-20(15-18)29-3;/h4-10,15,21H,11-14,16-17H2,1-3H3,(H,26,27);1H

InChI key

POQBIDFFYCYHOB-UHFFFAOYSA-N

General description

Sarpogrelate hydrochloride binds to the serotonin receptor (5-hydroxytryptamine, 5-HT2A). Its binding to 5-HT2A receptor inhibits thrombus formation and reduces platelet aggregation. Sarpogrelate hydrochloride has been used as an anti-atherosclerotic agent. It improves the endothelial function and may be useful for treating peripheral arterial disease. Use of sarpogrelate hydrochloride as 5-HT2A antagonist reduces urinary albumin levels in diabetic nephropathy.

Biochem/physiol Actions

Sarapogrelate hydrochloride is a potent and selective 5-HT2A receptor antagonist. It exhibits analgesic effects in numerous animal studies.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Duerschmied et al.
PloS one, 7(2), e32656-e32656 (2012-03-03)
Increased residual platelet reactivity remains a burden for coronary artery disease (CAD) patients who received a coronary stent and do not respond sufficiently to treatment with acetylsalicylic acid and clopidogrel. We hypothesized that serotonin antagonism reduces high on-treatment platelet reactivity.
Sarpogrelate hydrochloride, a selective 5-HT2A antagonist, improves vascular function in patients with peripheral arterial disease
Miyazaki M, et al.
Journal of Cardiovascular Pharmacology, 49(4), 221-227 (2007)
Shaoxin Ma et al.
Journal of diabetes investigation, 7(6), 833-844 (2016-11-03)
Our previous study found that dexamethasone-induced insulin resistance (IR) was involved in 5-hydroxytryptamine (5-HT) synthesis and 5-hydroxytryptamine 2 receptor (5-HT Male rats were exposed to dexamethasone for 10 days, then treated with or without a 5-HT Dexamethasone-induced whole-body IR, with
Sarpogrelate hydrochloride, a serotonin2A receptor antagonist, reduces albuminuria in diabetic patients with early-stage diabetic nephropathy
Takahashi T, et al.
Diabetes Research and Clinical Practice, 58(2), 123-129 (2002)
Identification of a key amino acid of the human 5-HT2B serotonin receptor important for sarpogrelate binding
Muntasir HA, et al.
Journal of Pharmacological Sciences, 104(3), 274-277 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service