Skip to Content
Merck
All Photos(1)

Key Documents

S5442

Sigma-Aldrich

SyntheChol® NS0 Supplement

500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture

Synonym(s):

Cholesterol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.75

biological source

non-animal source

sterility

sterile-filtered

product line

BioXtra

form

aqueous solution

concentration

500 ×

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

SMILES string

[H][C@@]12CC=C3C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])[C@H](C)CCCC(C)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

SyntheChol® NS0 Supplement is used as media supplement for NS0 mouse myeloma cell growth and the growth of other cholesterol-dependent cell lines.

Features and Benefits

Synthetic, non-animal cholesterol solution that is designed as a supplement to our hybridoma media formulations to promote excellent cell growth and protein productivity for NS0 mouse myeloma and other cholesterol-dependent cell lines.

Legal Information

SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur Nathan Brodsky et al.
Methods (San Diego, Calif.), 56(3), 408-417 (2011-10-29)
DNA microarray analysis of gene expression has become a valuable tool for bioprocessing research aimed at improving therapeutic protein yields. The highly parallel nature of DNA microarray technology allows researchers to assess hundreds of gene simultaneously, essentially enabling genome-wide snapshots.
Suba Dharshanan et al.
Cytotechnology, 66(4), 625-633 (2013-07-25)
From our recent publications, it was found that the deimmunization method (Dharshanan et al. (2012) Sci Res Essays 7:2288-2299) should be applied for the development of humanized anti-C2 monoclonal antibody (H1C2 mAb). However, the overlapping-PCR mutagenesis procedure used to insert
Marco Guadagnoli et al.
Blood, 117(25), 6856-6865 (2011-05-06)
APRIL (A proliferation-inducing ligand) is a TNF family member that binds two TNF receptor family members, TACI and BCMA. It shares these receptors with the closely related TNF family member, B-cell activating factor (BAFF). Contrary to BAFF, APRIL binds heparan
Steve Schoonooghe et al.
BMC biotechnology, 9, 70-70 (2009-08-13)
Tumour associated antigens on the surface of tumour cells, such as MUC1, are being used as specific antibody targets for immunotherapy of human malignancies. In order to address the poor penetration of full sized monoclonal antibodies in tumours, intermediate sized
Nancy Lynn Booth et al.
Cancer immunology, immunotherapy : CII, 58(8), 1229-1244 (2008-12-18)
We have developed a high-throughput screen (HTS) to search for novel molecules that can synergize with TRAIL, thus promoting apoptosis of ACHN renal tumor cells in a combinatorial fashion. The HTS detects synthetic compounds and pure natural products that can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service