Skip to Content
Merck
All Photos(3)

Key Documents

S3504

Sigma-Aldrich

Sigmacell Cellulose

Type 20, 20 μm

Synonym(s):

Cellulose, Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.56

type

Type 20

particle size

20 μm

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High purity cellulose powders for partition chromatography.
Makes extremely durable layers. Sharp separations for amino acids, sugars and other hydrophilic substances. Development time as short as 2 hours for 8′′ plates.
Sigmacell cellulose is used in thin layer chromatography (TLC) and cellulose TLC adsorbents. Sigmacell cellulose has been used in studies to inform industrial applications for large-scale purification of enzymes as well as to investigate the cellulase system of Penicillium echinulatum, which has been identified as a potential cellulase producer for bioconversion processes.

Reconstitution

For TLC, simply blend a 15-20% aqueous slurry for about one minute and coat plates; dry at room temperature. Usually does not require activation before use.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart M Linton et al.
The Journal of experimental biology, 207(Pt 23), 4095-4104 (2004-10-23)
Digestive juice from the herbivorous gecarcinid land crabs Gecarcoidea natalis and Discoplax hirtipes exhibited total cellulase activity and activities of two cellulase enzymes; endo-beta-1,4-glucanase and beta-1,4-glucosidase. These enzymes hydrolysed native cellulose to glucose. The digestive juice of both species also
Martin Bringmann et al.
Trends in plant science, 17(11), 666-674 (2012-07-13)
Directed plant cell growth is governed by deposition and alterations of cell wall components under turgor pressure. A key regulatory element of anisotropic growth, and hence cell shape, is the directional deposition of cellulose microfibrils. The microfibrils are synthesized by
Olof Regnell et al.
Ambio, 43(7), 878-890 (2014-01-15)
Methylation of mercury (Hg) to highly toxic methyl Hg (MeHg), a process known to occur when organic matter (OM) decomposition leads to anoxia, is considered a worldwide threat to aquatic ecosystems and human health. We measured temporal and spatial variations
Okako Omadjela et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(44), 17856-17861 (2013-10-16)
Cellulose is a linear extracellular polysaccharide. It is synthesized by membrane-embedded glycosyltransferases that processively polymerize UDP-activated glucose. Polymer synthesis is coupled to membrane translocation through a channel formed by the cellulose synthase. Although eukaryotic cellulose synthases function in macromolecular complexes
Nathalie Lavoine et al.
Carbohydrate polymers, 90(2), 735-764 (2012-07-31)
Interest in microfibrillated cellulose (MFC) has been increasing exponentially. During the last decade, this bio-based nanomaterial was essentially used in nanocomposites for its reinforcement property. Its nano-scale dimensions and its ability to form a strong entangled nanoporous network, however, have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service