Skip to Content
Merck
All Photos(1)

Key Documents

S300HR

Sigma-Aldrich

Sephacryl® Size Exclusion Resin

300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)

Synonym(s):

Poly([allyl dextran]-co-N,N′-methylenebisacrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

product name

Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)

Quality Level

description

stability: stable to 0.2M NaOH

form

suspension (20% aqueous ethanol)

feature

autoclavable

matrix

crosslinked allyl dextran/N,N′-methylenebisacrylamide copolymer

particle size

25-75 μm (wet)

pore size

1-400 kDa MW range (dextrans)
10-1500 kDa MW range (globular proteins)

compatibility

mode of use size exclusion chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

S300HR-750ML′s updated product number is GE17-0599-01
  • Supplied swollen, suspended in 20% aqueous ethanol
  • Autoclavable
  • Stable to 0.2N sodium hydroxide (NaOH)
  • Store at refrigerated temperature (2-8 °C)

Application

Sephacryl® is used in gel filtration chromatography, protein chromatography, and resins and separation media. Sephacryl® has been used to develop a novel high-throughput screening assay for sickle cell disease drug discovery. Sephacryl® has also been used to purify and characterize ovine pancreatic elastase as well as a lysozyme from human urine.
For size exclusion chromatography; the HR grade is a smaller particle size with narrower size range distribution, allowing for more efficient separations at faster flow rates. Recommended fractionation ranges are listed for globular proteins and dextrans..

Legal Information

Sephacryl is a registered trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F - closed cup

Flash Point(C)

38 - 43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Soos et al.
The Biochemical journal, 263(2), 553-563 (1989-10-15)
The receptors for insulin and insulin-like growth factor-I (IGF-I) are closely related in primary sequence and overall structure. We have examined the immunological relationships between these receptors by testing the reactivity of anti-(insulin receptor) monoclonal antibodies with IGF-I receptors in
Danielle Clérot et al.
Journal of virology, 80(22), 11322-11330 (2006-09-01)
The Rep protein of tomato yellow leaf curl Sardinia virus (TYLCSV), a single-stranded DNA virus of plants, is the replication initiator essential for virus replication. TYLCSV Rep has been classified among ATPases associated with various cellular activities (AAA+ ATPases), in
Hargeet K Brar et al.
Molecular plant-microbe interactions : MPMI, 24(10), 1179-1188 (2011-06-04)
Fusarium virguliforme causes sudden death syndrome (SDS) in soybean. The pathogen has never been isolated from diseased foliar tissues; therefore, one or more toxins have been considered to cause foliar SDS development. Cell-free F. virguliforme culture filtrates containing a toxin
J M Gibbins et al.
The Journal of biological chemistry, 273(51), 34437-34443 (1998-12-16)
There is extensive evidence to show that phosphatidylinositol 3-kinase plays an important role in signaling by the immune family of receptors, which has recently been extended to include the platelet collagen receptor, glycoprotein VI. In this report we present two
S H Zaidi et al.
The Journal of biological chemistry, 270(29), 17292-17298 (1995-07-21)
The central nervous system deposition by neurons and glia of beta A4 amyloid protein is an important contributing factor to the development of Alzheimer's disease. Amyloidogenic cells overexpress amyloid precursor protein (APP) mRNAs suggesting a transcriptional or post-transcriptional defect may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service