Skip to Content
Merck
All Photos(1)

Key Documents

S1198

Sigma-Aldrich

Sumatriptan succinate

≥98% (HPLC), solid, 5-HT₁ receptor agonist

Synonym(s):

3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide succinate, GR-43175

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O2S · C4H6O4
CAS Number:
Molecular Weight:
413.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Sumatriptan succinate, ≥98% (HPLC), solid

Assay

≥98% (HPLC)

form

solid

color

white to beige

solubility

H2O: >20 mg/mL

originator

GlaxoSmithKline

SMILES string

OC(=O)CCC(O)=O.CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)C)c2c1

InChI

1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8)

InChI key

PORMUFZNYQJOEI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sumatriptan succinate has the ability to constrict human cranial arteries. This 5-HT1 receptor agonist can be used to treat migraine.
Sumatriptan succinate is a 5-HT1 serotonin receptor agonist.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Treatment of acute migraine with subcutaneous sumatriptan
Cady RK, et al.
JAMA : The Journal of the American Medical Association, 265(21), 2831-2835 (1991)
Mucoadhesive bilayered patches for administration of sumatriptan succinate
Shidhaye SS, et al.
Aaps Pharmscitech, 9(3), 909-916 (2008)
Tse-Ming Chou et al.
The journal of headache and pain, 23(1), 157-157 (2022-12-13)
To investigate specific brain regions and neural circuits that are responsible for migraine chronification. We established a mouse model of chronic migraine with intermittent injections of clinically-relevant dose of nitroglycerin (0.1 mg/kg for 9 days) and validated the model with cephalic and
Toshihiko Tomita et al.
Journal of gastroenterology and hepatology, 28(1), 106-111 (2012-09-20)
Scintigraphy is a useful noninvasive technique for assessment of gastric motility, especially emptying, but there is little knowledge of use of the technique to assess gastric accommodation. Therefore, to clarify the usefulness of scintigraphy as a technique for assessing gastric
Benjamin W Friedman et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 19(10), 1151-1157 (2012-09-22)
Patients who use an emergency department (ED) for acute migraine headaches have higher migraine disability scores, lower socioeconomic status, and are unlikely to have used a migraine-specific medication prior to presentation to the ED. The objective was to determine if

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service