S1020
Screens for CD-1™
size 60 mesh
Synonym(s):
cell sieve, mesh screen, screen mesh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
size
60 mesh
Looking for similar products? Visit Product Comparison Guide
General description
Screens for CD-1™ is a stainless-steel celldissociation sieve designed for the preparation of single-cell suspensions fromtissues and fragment polyacrylamide gels.
Application
Screens for CD-1™ have been used for:
- filtering tracheids, wood fibers, and ray parenchyma cells from hydrogen peroxide-acetic acid (HPAC)-pretreated oak
- in the isolation of enterocytes from Oncorhynchus mykiss intestine
- to filter dissociated chicken testicular cells to obtain single-cell suspension
Legal Information
CD-1 is a trademark of Sigma-Aldrich Co. LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecular evidence and physiological characterization of iron absorption in isolated enterocytes of rainbow trout (Oncorhynchus mykiss): Implications for dietary cadmium and lead absorption
Aquatic Toxicology (Amsterdam, Netherlands), 343-350, 99(13)-99(13) (2010)
Formation of testicular structure under skin after auto-transplantation of dispersed testicular cells in the chicken
Animal Reproduction Science, 120, 125-128 (2010)
Biotechnology for biofuels, 14(1), 37-37 (2021-02-08)
Woody plants with high glucose content are alternative bioresources for the production of biofuels and biochemicals. Various pretreatment methods may be used to reduce the effects of retardation factors such as lignin interference and cellulose structural recalcitrance on the degradation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service