Skip to Content
Merck
All Photos(1)

Key Documents

S0292

Sigma-Aldrich

Silymarin

flavonolignans

Synonym(s):

flavolignans

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.81

biological source

plant (Silybum marianum)

Quality Level

form

powder

technique(s)

toxicology assay: suitable

application(s)

microbiology

storage temp.

−20°C

SMILES string

O1C(C(Oc3c1ccc(c3)C4Oc5c(c(cc(c5)O)O)C(=O)C4O)c2cc(c(cc2)O)OC)CO

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3

InChI key

SEBFKMXJBCUCAI-UHFFFAOYSA-N

General description

Silymarin is a flavonolignan, obtained from milk thistle (Silybum marianum) plant.

Application

Silymarin has been used to study:
  • its in vitro antiviral, antibacterial, antifungal activities and cytotoxicity
  • its effect of silymarin on bladder contractions in cyclophosphamide (CYP)-induced cystitis rat model
  • its effect on liver toxication induced by Fumonisin B1 in mice

Biochem/physiol Actions

Silymarin is originally known to protect liver from various toxic agents and is used in treating hepatitis and cirrhosis. Silymarin possesses many biological actions such as antifibrotic, anti-lipid peroxidative, anti-inflammatory, immunomodulatory and membrane stabilizing. Silymarin also aids in liver regeneration.
Silymarin provides cardioprotective activity against ischemia-reperfusion induced myocardial infarction in rats.
Silymarin was shown to protect the liver from the cytotoxic effects of anti-tuberculosis drugs by decreasing serum alanine aminotransferase (ALT), aspartate aminotransferase (AST) and alkaline phosphatase (ALP) levels. This effect was related to the anti-oxidant effects of silymarin.

Other Notes

Mixture of anti-hepatotoxic flavonolignans from the fruit of Silybum marianum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vieri Piazzini et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-06)
Two novel nanomicellar formulations were developed to improve the poor aqueous solubility and the oral absorption of silymarin. Polymeric nanomicelles made of Soluplus and mixed nanomicelles combining Soluplus with d-α-tocopherol polyethylene glycol 1000 succinate (vitamin E TPGS) were prepared using
Effect of silymarin on bladder overactivity in cyclophosphamide-induced cystitis rat model
Eser N, et al.
Phytomedicine, 19(8-9), 840-845 (2012)
Sirajudheen Anwar et al.
Indian journal of pharmacology, 50(3), 108-115 (2018-09-01)
Silymarin, extracted from the seeds of Silybum marianum L. (Milk thistle), is traditionally used for treating various illnesses such as diabetes, cancer, inflammation, hepatitis, liver cirrhosis, and renal problems. Acute cytotoxicity and genotoxicity studies have been reported with ambiguous outcomes;
Laise Mara Oliveira Miranda et al.
Biomedical and environmental sciences : BES, 33(9), 690-700 (2020-10-28)
To evaluate the efficiency of silymarin (SMN) in modulating metabolic parameters and redox status in rats with type 1 diabetes mellitus (T1DM). Diabetes was induced by intraperitoneal injection of alloxan. The diabetic rats were administered with SMN at doses of
Chun-Ping Lu et al.
Chemico-biological interactions, 290, 12-18 (2018-05-14)
Macrophages and inflammasome pathway are involved in high-glucose toxicity and development of insulin resistance. Silymarin (SMR) was known to modulate glucose homeostasis and reduce inflammation. However, it is still unknown whether SMR possess anti-hyperglycemic effects in diabetic-like knockout mice (Hnf-1αkin/-/Ins.cre

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service