Skip to Content
Merck
All Photos(2)

Key Documents

RS2

Sigma-Aldrich

Rinse Solution no. 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

Quality Level

IVD

for in vitro diagnostic use

concentration

1.6 % (w/v)

application(s)

hematology
histology

storage temp.

room temp

Application

Intended for use as a buffer with various Romanowky type staining procedures, specifically those performed on the Hematek. RS2 is a component of Sigma′s Wright Stain Set (Cat. No. WSHT-1SET).

Components

Ethanol solution, 18% w/v, with surfactant. Sodium azide, 0.02% added as preservative.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jajati K Mohapatra et al.
The Journal of general virology, 96(Pt 3), 553-564 (2014-11-09)
In this study we describe the adaptive changes fixed on the capsid of several foot-and-mouth disease virus serotype A strains during propagation in cell monolayers. Viruses passaged extensively in three cell lines (BHK-21, LFBK and IB-RS-2) consistently gained positively charged
Tovhowani D Ramulongo et al.
Veterinary microbiology, 243, 108614-108614 (2020-04-11)
Foot-and-mouth disease (FMD) is a highly contagious vesicular disease of cloven-hoofed animals, which severely decreases livestock productivity. FMD virus (FMDV), the causative agent, initiates infection by interaction with integrin cellular receptors on pharyngeal epithelium cells, causing clinical signs one to
M Chitray et al.
Archives of virology, 159(5), 947-961 (2013-11-14)
Genetic information regarding the leader (L) and complete capsid-coding (P1) region of FMD serotype A and O viruses prevalent on the African continent is lacking. Here, we present the complete L-P1 sequences for eight serotype A and nine serotype O
Katherine A Scott et al.
Vaccine, 35(40), 5426-5433 (2017-04-23)
Foot-and-mouth disease (FMD) vaccines with improved stability and less reliant on a cold-chain are needed to improve the longevity of immune responses elicited in animals. This is especially so for serotypes O and SAT2 which are unstable in mildly acidic
Dong-Hyun Jung et al.
3 Biotech, 10(2), 31-31 (2020-01-29)
Resistant starch (RS) is an important food source from which gut bacteria produce short chain fatty acids, which have beneficial effects for human health. The Bifidobacterium adolescentis P2P3, a human gut bacterium possessing a strong RS-degrading activity, was isolated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service