RFA452411-EP
Rotor for Eppendorf® 5430/5430R Centrifuges
F-45-24-11, non aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid: no
All Photos(2)
About This Item
UNSPSC Code:
41103909
Recommended Products
feature
lid: no
to hold 24 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639471 (NA)
Eppendorf® 5702746002 (EU)
parameter
1,770 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)