Skip to Content
Merck
All Photos(5)

Key Documents

RDD021

Sigma-Aldrich

Sodium dodecyl sulfate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

description

anionic

product line

Redi-Dri

Assay

≥99.0%

quality

free-flowing

mol wt

288.38 g/mol

impurities

≤0.06 meq/g Titr. Base
≤4.0% unsulfated alcohols
≥96.0% fatty alcohols

loss

≤1.0% loss on drying

mp

204-207 °C (lit.)

solubility

water: soluble 100 mg/mL (opalescent solution)

cation traces

heavy metals (as Pb): ≤0.002%

λ

3 %H2O reference

UV absorption

λ: 230-350 nm Amax: ≤0.1

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

SDS (sodium dodecyl sulfate) is a wetting agent and an anionic detergent which functions efficiently in both alkaline and acidic solutions. It is generally used for the solubilization of lipids and proteins. It also functions as a potent protein denaturant. SDS, volume-wise, is one of the most commonly used synthetic surfactants.

Application

Anionic detergent

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicity of anionic detergents determined by Saccharomyces cerevisiae microarray analysis.
Sirisattha S et al
Water Research, 38(1), 61-70 (2004)
M D Womack et al.
Biochimica et biophysica acta, 733(2), 210-215 (1983-09-07)
Over 50 detergents were tested to establish which would be most effective in releasing proteins from membrane-bounded compartments without denaturing them. Various concentrations each of detergent were tested for two activities: (1) solubilization of egg phospholipid liposomes as measured by
Antony Anet et al.
Journal of hazardous materials, 370, 42-53 (2018-09-15)
This study investigates Bisphenol A (BPA) induced oxidative stress that mediates the genotoxicity in in vivo model Drosophila melanogaster. The calculated LC50 for BPA was 12.35 μg/mL. The strains of D. melanogaster were reared in 0.1, 1.0, 2.5 and 5.0 μg/mL BPA
Erdem D Tabdanov et al.
Cell reports, 25(2), 328-338 (2018-10-12)
Cancer cell migration through and away from tumors is driven in part by migration along aligned extracellular matrix, a process known as contact guidance (CG). To concurrently study the influence of architectural and mechanical regulators of CG sensing, we developed
Daniel Zingg et al.
Cancer cell, 34(1), 69-84 (2018-07-17)
Human melanomas frequently harbor amplifications of EZH2. However, the contribution of EZH2 to melanoma formation has remained elusive. Taking advantage of murine melanoma models, we show that EZH2 drives tumorigenesis from benign BrafV600E- or NrasQ61K-expressing melanocytes by silencing of genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service