RAB0764
Mouse C5a / Complement C5a ELISA Kit
for serum, plasma and cell culture supernatants
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 1.64-400 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... C5(15139)
General description
The antibody pair provided in this kit recognizes mouse Complement Component C5a.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frédéric Lavainne et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(6), 1225-1231 (2014-02-12)
Soluble Flt1 (sFlt1) is a potent inhibitor of vascular endothelial growth factor, secreted mainly by the placenta, endothelial cells and monocytes. Increased sFlt1 serum levels correlate with endothelial dysfunction and cardiovascular complications in dialysis patients. However, the impact of dialysis
Cynthia Owsley et al.
Investigative ophthalmology & visual science, 55(8), 4776-4789 (2014-05-24)
Delayed rod-mediated dark adaptation (DA) is characteristic of early age-related macular degeneration (AMD) and also can be observed in some older adults in normal macular health. We examine cross-sectional associations between rod-mediated DA and risk factors for AMD in older
Michael T Eadon et al.
Immunology, 143(3), 363-373 (2014-05-08)
Blood-brain barrier (BBB) disturbance is a crucial occurrence in many neurological diseases, including systemic lupus erythematosus (SLE). Our previous studies showed that experimental lupus serum altered the integrity of the mouse brain endothelial layer, an important constituent of the BBB.
Huifang Tang et al.
Journal of immunology (Baltimore, Md. : 1950), 193(7), 3769-3778 (2014-08-31)
Increasing evidence suggests that the novel anti-inflammatory and proresolving mediators such as the resolvins play an important role during inflammation. However, the functions of these lipid mediators in immune complex-induced lung injury remain unknown. In this study, we determined the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service