Skip to Content
Merck
All Photos(1)

Key Documents

RAB0642

Sigma-Aldrich

Human F3 / Tissue Factor ELISA Kit

Synonym(s):

Coagulation factor III, TF, Thromboplastin, Tissue factor 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... F3(2152)

General description

The antibody pair provided in this kit recognizes human Coagulation Factor III/Tissue Factor.

Tissue factor (TF), also known as coagulation factor III (F3), is encoded by the gene mapped to human chromosome 1p21.3. TF has been shown to exist in two forms, namely membrane-bound and soluble TF. The encoded protein contains an extracellular domain (ED), a single transmembrane domain (TD) and short cytoplasmic domain (CD).

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Human F3 / Tissue Factor ELISA Kit has been used to measure the concentration of soluble tissue factor (TF) expressed in human umbilical vein smooth muscle cells (HUVSMCs).

Biochem/physiol Actions

Tissue factor (TF) interacts with the zymogen factor VII (VII) and its activated form VIIa, and initiates blood coagulation protease cascade. The encoded protein is involved in the pathogenesis of cardiovascular diseases, such as hypertension, atherosclerosis, acute coronary syndrome and restenosis. Thus, it is considered to be a potential therapeutic target for cardiovascular diseases.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saika Manzoor et al.
Scientific reports, 9(1), 10182-10182 (2019-07-17)
Polycystic ovarian syndrome (PCOS) is a multispectral disorder requiring lifelong management. Its pathophysiology is still being explored which makes its treatment options restrained. Present study explores impact of oral contraceptive mode of treatment on metabolic, hormonal, inflammation and coagulation profile
Bin Jiang et al.
Biotechnology and bioengineering, 114(10), 2371-2378 (2017-05-26)
The clinical translation of cell-based therapeutics often requires highly sensitive, non-invasive imaging tools to assess cell function and distribution in vivo. The objective of this research was to determine whether human Sodium-Iodide Symporter (hNIS) ectopic expression in endothelial cells (ECs)
Cytogenetics and molecular genetics of prostate cancer: A comprehensive update.
Fonseka L N, et al.
Journal of the Association of Genetic Technologists, 41, 100-111 (2015)
Alberto Centeno et al.
Journal of investigative surgery : the official journal of the Academy of Surgical Research, 33(4), 339-349 (2018-11-01)
Background: Excessive blood loss is a relevant complication of partial liver resection. Topical hemostatic agents have proven useful to improve the control of the bleeding in this among other surgical indications. Until now all of these products have been based
J Bartko et al.
Journal of thrombosis and haemostasis : JTH, 14(12), 2471-2477 (2016-10-25)
Essentials Glucocorticoids are associated with an increased risk of thrombosis. Healthy volunteers received dexamethasone or placebo in an endotoxin lung instillation model. Dexamethasone suppressed thrombin generation in bronchoalveolar lavage. Glucocorticoids inhibit endotoxin induced pulmonary coagulopathy. Background Activation of local and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service