RAB0541
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit
for serum, plasma, cell culture supernatants and urine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
sample type urine
assay range
inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.6 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IFNA1(3439)
General description
This ELISA antibody pair detects human Interferon Alpha-1/13.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Satu Wedenoja et al.
EBioMedicine, 59, 102872-102872 (2020-07-19)
Fetal immune tolerance is crucial for pregnancy success. We studied the link between preeclampsia, a severe pregnancy disorder with uncertain pathogenesis, and fetal human leukocyte antigen G (HLA-G) and other genes regulating maternal immune responses. We assessed sex ratios and
Tauyne Menegaldo Pinheiro et al.
PLoS neglected tropical diseases, 12(5), e0006525-e0006525 (2018-05-31)
The dynamics of dengue virus (DENV) circulation depends on serotype, genotype and lineage replacement and turnover. In São José do Rio Preto, Brazil, we observed that the L6 lineage of DENV-1 (genotype V) remained the dominant circulating lineage even after
Vibha Jha et al.
PloS one, 9(8), e104484-e104484 (2014-08-15)
Environmental factors including drugs, mineral oils and heavy metals such as lead, gold and mercury are triggers of autoimmune diseases in animal models or even in occupationally exposed humans. After exposure to subtoxic levels of mercury (Hg), genetically susceptible strains
Shigeo Koido et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(16), 4228-4239 (2014-07-25)
We performed a phase I trial to investigate the safety, clinical responses, and Wilms' tumor 1 (WT1)-specific immune responses following treatment with dendritic cells (DC) pulsed with a mixture of three types of WT1 peptides, including both MHC class I
Liang-Chuan S Wang et al.
Cancer immunology research, 2(2), 154-166 (2014-04-30)
The majority of chimeric antigen receptor (CAR) T-cell research has focused on attacking cancer cells. Here, we show that targeting the tumor-promoting, nontransformed stromal cells using CAR T cells may offer several advantages. We developed a retroviral CAR construct specific
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service