RAB0314
Mouse IL-6 R ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Il6Ra(16194)
General description
The Mouse IL-6R (interleukin 6 soluble receptor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-6R in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse IL-6 R
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Christian König et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(38), 9782-9791 (2016-09-23)
During peripheral inflammation, both spinal TNF-α and IL-6 are released within the spinal cord and support the generation of inflammation-evoked spinal hyperexcitability. However, whether spinal TNF-α and IL-6 act independently in parallel or in a functionally dependent manner has not
Christoffer K Goth et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(47), 14623-14628 (2015-11-11)
Regulated shedding of the ectodomain of cell membrane proteins by proteases is a common process that releases the extracellular domain from the cell and activates cell signaling. Ectodomain shedding occurs in the immediate extracellular juxtamembrane region, which is also where
Jean Gaschignard et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 59(2), 244-251 (2014-04-25)
About 10% of pediatric patients with invasive pneumococcal disease (IPD) die from the disease. Some primary immunodeficiencies (PIDs) are known to confer predisposition to IPD. However, a systematic search for these PIDs has never been carried out in children presenting
K Hultman et al.
Journal of thrombosis and haemostasis : JTH, 12(5), 701-712 (2014-03-13)
Excess fibrin in blood vessels is cleared by plasmin, the key proteolytic enzyme in fibrinolysis. Neurological disorders and head trauma can result in the disruption of the neurovasculature and the entry of fibrin and other blood components into the brain
Jin Wang et al.
Molecular medicine reports, 24(5) (2021-09-04)
Ischemic stroke is one of the most common diseases that has a high rate of mortality, and has become a burden to the healthcare system. Previous research has shown that EPH receptor B4 (EphB4) promotes neural stem cell proliferation and differentiation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service