Skip to Content
Merck
All Photos(3)

Documents

R7752

Sigma-Aldrich

Ristomycin monosulfate

≥90% ristocetin A basis (balance primarily Ristocetin B), powder, platelet activator

Synonym(s):

Ristocetin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C95H110N8O44 · H2SO4
CAS Number:
Molecular Weight:
2166.00
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Ristomycin monosulfate, ≥90% ristocetin A basis (balance primarily Ristocetin B)

Quality Level

Assay

≥90% ristocetin A basis (balance primarily Ristocetin B)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H]1NC(=O)[C@H]2NC(=O)C(NC(=O)[C@@H]3NC(=O)[C@H]4NC(=O)[C@H](NC(=O)[C@H](N)c5ccc(O)c(Oc6cc4cc(O)c6C)c5)[C@H](O)c7ccc(Oc8cc3cc(Oc9ccc(cc9)[C@H]2O[C@H]%10C[C@@H](N)[C@@H](O)[C@H](C)O%10)c8O[C@@H]%11O[C@H](CO[C@@H]%12O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%12O)[C@@H](O)[C@H](O)[C@H]%11O[C@H]%13O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]%13O[C@@H]%14OC[C@@H](O)[C@@H](O)[C@@H]%14O)cc7)c%15ccc(O)c(c%15)-c%16c(O[C@@H]%17O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]%17O)cc(O)cc1%16

InChI

1S/C95H110N8O44/c1-30-47(109)18-37-20-49(30)139-50-19-35(9-16-46(50)108)59(97)84(125)102-64-68(113)33-5-11-40(12-6-33)137-52-21-38-22-53(81(52)145-95-83(76(121)72(117)56(143-95)29-134-91-78(123)73(118)67(112)32(3)136-91)147-94-82(75(120)71(116)55(27-105)142-94)146-92-77(122)69(114)48(110)28-133-92)138-41-13-7-34(8-14-41)80(144-57-25-44(96)66(111)31(2)135-57)65-89(130)101-63(90(131)132-4)43-23-39(106)24-51(140-93-79(124)74(119)70(115)54(26-104)141-93)58(43)42-17-36(10-15-45(42)107)60(85(126)103-65)98-87(128)62(38)99-86(127)61(37)100-88(64)129/h5-24,31-32,44,48,54-57,59-80,82-83,91-95,104-124H,25-29,96-97H2,1-4H3,(H,98,128)(H,99,127)(H,100,129)(H,101,130)(H,102,125)(H,103,126)/t31-,32-,44+,48+,54+,55+,56+,57-,59+,60?,61-,62+,63?,64+,65-,66-,67-,68+,69+,70+,71+,72+,73+,74-,75-,76-,77-,78+,79-,80+,82-,83+,91+,92-,93+,94+,95-/m0/s1

InChI key

VUOPFFVAZTUEGW-ZNAZZOOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ristomycin/ristocetin is a type III glycopeptide. It is a highly glycosylated glycopeptide, previously found in Amycolatopsis lurida.

Application

Ristomycin monosulfate has been used:
  • to activate platelet and induce platelet aggregation
  • to study the aggregation responses of platelets from buffalo, horse, pig and sheep
  • in in-house ristocetin cofactor von Willebrand factor (VWF:RCo) assay to measure glycoprotein Ib (GpIb) binding activity

Biochem/physiol Actions

Ristocetin activates platelets. Ristocetin-induced platelet agglutination (RIPA) is decreased in patients undergoing chronic hemodialysis. Together with increased plasma glycocalicin levels (a product of enzymatic cleavage of GPIb), RIPA may contribute to diminished platelet adhesion to vascular subendothelium and increased bleeding associated with uremia. Addition of ristocetin and vWf caused specific agglutination of rGPIbα-liposomes suporting the potential use of rGPIbα-liposomes to enhance platelet function in vivo and support hemostasis in thrombocytopenic individuals.
Ristomycin possesses antibacterial activity. It may be used to diagnose von Willebrand disease and Bernard-Soulier syndrome. Ristocetin-induced platelet agglutination (RIPA) is decreased in patients undergoing chronic hemodialysis. Increased plasma glycocalicin levels (a product of enzymatic cleavage of GPIb) and decreased RIPA might reduce platelet adhesion to vascular subendothelium and improve bleeding associated with uremia in patients with chronic hemodialysis.

Other Notes

Glycopeptide antibiotic complex

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordan, D.C. et al.
Antibiotics, 1, 84-84 (1967)
Bleeding Symptoms and von Willebrand Factor Levels: 30-Year Experience in a Tertiary Care Center
Moonla C, et al.
Clinical and Applied Thrombosis/Hemostasis : Official Journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis (2019)
J Himmelfarb et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 32(1), 132-138 (1998-07-21)
A bleeding diathesis caused by platelet dysfunction is a major cause of morbidity and mortality in patients with uremia. Platelet adhesion to vascular subendothelium is defective in uremia and depends on the interactions of the platelet glycoprotein (GP) Ib/IX complex
Novel antibodies against GPIb alpha inhibit pulmonary metastasis by affecting vWF-GPIb alpha interaction
Qi Y, et al.
Journal of Hematology & Oncology, 11(1), 117-117 (2018)
T Kitaguchi et al.
Biochemical and biophysical research communications, 261(3), 784-789 (1999-08-12)
Platelet glycoprotein (GP) Ib/IX/V complex is a receptor for von Willebrand factor (vWf), which plays a crucial role in primary hemostasis by mediating platelet adhesion to injured blood vessels. We have expressed in CHO cells a fragment of GPIba that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service