R5130
Anti-Rat IgG (whole molecule) antibody produced in goat
affinity isolated antibody, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
lyophilized powder
technique(s)
quantitative precipitin assay: suitable
storage temp.
2-8°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Binds all rat Igs.
Application
Anti-Rat IgG (whole molecule) antibody produced in goat was used for immunoglobulin quantification by ELISA at a dilution of 1:500-1:1000. It was used in gel supershift analysis to study DNA-binding activity of PPARα.
Biochem/physiol Actions
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Physical form
Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2
Reconstitution
Reconstitute with 0.135 M sodium chloride
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hendry Susila et al.
Plant communications, 4(3), 100515-100515 (2023-01-05)
The timing of flowering is tightly controlled by signals that integrate environmental and endogenous cues. Sugars produced by carbon fixation in the chloroplast are a crucial endogenous cue for floral initiation. Chloroplasts also convey information directly to the nucleus through
A Benani et al.
Neuroscience letters, 369(1), 59-63 (2004-09-24)
Following recurrent noxious stimulation, both functional modification and structural reorganization such as activation of the arachidonate cascade or axon sprouting occur in the central nervous system (CNS). It has been recently proposed that these alterations observed during chronic pain state
Rong Deng et al.
Blood, 109(6), 2470-2476 (2006-11-30)
The present work evaluated antibody-coated liposomes as a new treatment strategy for immune thrombocytopenic purpura (ITP) through the use of a mouse model of the disease. Effects of antimethotrexate antibody (AMI)-coated liposomes and intravenous immunoglobulin (IVIG)-coated liposomes (15, 30, 60
Zijie Zhou et al.
Biology, 10(12) (2021-12-25)
Nonhuman primates (NHPs) are relevant models to study the pathogenesis of tuberculosis (TB) and evaluate the potential of TB therapies, but rapid tools allowing diagnosis of active pulmonary TB in NHPs are lacking. This study investigates whether low complexity lateral
S Nutten et al.
Clinical and vaccine immunology : CVI, 14(2), 190-197 (2006-12-08)
In this study, we assessed the effect of administering the antibiotic amoxicillin to rat pups on the immune response to orally fed ovalbumin (OVA). We first established that amoxicillin administration durably altered the gut microbiota of these animals. In parallel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service