Skip to Content
Merck
All Photos(2)

Key Documents

R4779

Sigma-Aldrich

Anti-Rab7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ras-Related Protein Rab7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

mouse, human, rat

technique(s)

indirect immunofluorescence: 5-10 μg/mL using rat NRK and mouse NIH-3T3 cells fixed and permeabilized with 3% paraformaldehyde followed by 0.4% saponin
western blot: 2.5-5.0 μg/mL using extract of human A431 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB7A(7879)
mouse ... Rab7(19349)
rat ... Rab7a(29448)

General description

Rab7 is a member of the Rab family of small guanosine triphosphatases (GTPases). Rab proteins contain conserved regions involved in guanine nucleotide binding, and hypervariable COOH-terminal domains with a cysteine motif, implicated in subcellular targeting. Each Rab protein shows a characteristic subcellular distribution. The antibody is affinity-purified using the immunizing peptide immobilized on agarose.
Ras-related protein Rab-7 (Rab7) is a member of the Rab family of small guanosine triphosphatases (GTPases).

Specificity

Anti- Rab7 recognizes human, mouse and rat Rab7.

Immunogen

Synthetic peptide corresponding to amino acid residues of human Rab7 with C-terminal added cysteine, conjugated to KLH.

Application

Anti-Rab7 antibody can be used in indirect immunofluorescence and immunoblotting.
Anti-Rab7 antibody produced in rabbit has been used in confocal microscopy.

Biochem/physiol Actions

Rab family of small guanosine triphosphatases (GTPases) are central regulators of membrane trafficking between the different subcellular compartments of the eukaryotic cell. Their regulatory capacity depends on their ability to cycle between the GDP-bound inactive and GTP-bound active states. Conversion from one state to the other is regulated by GDP/GTP exchange factors (GEPs), GDP dissociation inhibitors (GDIs) and GTPase-activating proteins (GAPs). Activation of a Rab protein is coupled to its association with intracellular membranes, allowing it to recruit downstream effector proteins to the cytoplasmic surface of a subcellular compartment. Through their effector proteins, Rab GTPases regulate vesicle formation, actin- and tubulin-dependent vesicle movement, and membrane fusion. Rab7 is also involved in the maturation of late autophagic vacuoles. Among Rab7 effectors are RILP (Rab-interacting lysosome protein) that controls late endosomal and lysosomal transport by mediating the recruitment of dynein/dynactin motors, Rabring7 (Rab7- interacting ring finger protein),(10) and the hVPS34/p150 complex. Anti-Rab7 may be used as a marker for late endosomes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human VPS34 and p150 are Rab7 interacting partners
Stein MP, et al.
Traffic, 4(11), 754-771 (2003)
IL-7 induces clathrin-mediated endocytosis of CD127 and subsequent degradation by the proteasome in primary human CD8 T cells
Faller EM, et al.
Immunology and Cell Biology, 94(2), 196-207 (2016)
Rab7 and Rab9 are recruited onto late endosomes by biochemically distinguishable processes
Soldati T, et al.
The Journal of Biological Chemistry, 270(43), 25541-25548 (1995)
Role for Rab7 in maturation of late autophagic vacuoles
Jager S, et al.
Journal of Cell Science, 117(20), 4837-4848 (2004)
Structural basis for recruitment of RILP by small GTPase Rab7
Wu Mousheng, et al.
The Embo Journal, 24(8), 1491-1501 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service