Skip to Content
Merck
All Photos(1)

Key Documents

R3405

Sigma-Aldrich

RP-107

≥98% (HPLC), solid

Synonym(s):

7-n-Butyl-6-(4-hydroxyphenyl)[5H]-pyrrolo[2,3-b]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O
CAS Number:
Molecular Weight:
267.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCCCc1c([nH]c2nccnc12)-c3ccc(O)cc3

InChI

1S/C16H17N3O/c1-2-3-4-13-14(11-5-7-12(20)8-6-11)19-16-15(13)17-9-10-18-16/h5-10,20H,2-4H2,1H3,(H,18,19)

InChI key

PRIGRJPRGZCFAS-UHFFFAOYSA-N

Biochem/physiol Actions

RP107 is an activator of cystic fibrosis transmembrane conductance regulator (CFTR) channel.
RP107 is an activator of cystic fibrosis transmembrane conductance regulator (CFTR) channel. RP107 stimulates wild-type CFTR and mutated CFTR with submicromolar affinity by a cAMP-independent mechanism. EC50 = 89 nM at activating CFTR-dependent C-secretion, an effect inhibited by CFTR(inh)-172 and glibenclaminde. RP107 constitutes a new example of a scaffold structure for the selective activation of CFTR receptors.

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service