Skip to Content
Merck
All Photos(1)

Key Documents

R2030

Sigma-Aldrich

Anti-RACK1 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution, immunoprecipitation: 5 μg

Synonym(s):

Anti-GNB2L1, Anti-Gnb2-rs1, Anti-Guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1, Anti-H12.3, Anti-HLC-7, Anti-Lung cancer oncogene 7, Anti-PIG21, Anti-Proliferation-inducing gene 21, Anti-Receptor of Activated Protein Kinase C1, Anti-protein homologous to chicken B complex protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg
western blot: 0.5-1 μg/mL using HEK-293T cell

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNB2L1(10399)
mouse ... Gnb2l1(14694)
rat ... Gnb2l1(83427)

General description

Receptors for activated C-kinase (RACKs) are scaffold proteins for protein kinase C (PKC). It is composed of seven WD repeats, thus resembling the structure of the β subunit of G proteins. It is not a substrate of PKC, but in its presence, the activity of PKC is increased by several fold.

Immunogen

synthetic peptide corresponding to amino acids 268-282 of human RACK1, conjugated to KLH via an N-terminal added cystein residue. The corresponding sequence is conserved in human, rat and mouse.

Biochem/physiol Actions

RACK1 is a receptor for activated C kinases that functions downstream of the growth factor receptor signaling pathway. RACK1 also modulates several functions such as cell adhesion, chemotaxis and protrusion. Increased RACK1 expression has been associated with cell migration and metastasis in breast carcinoma. Thus, RACK1 can be used as a prognostic indicator for breast cancer
Receptors for activated C-kinase (RACKs) anchor activated protein kinase C (PKC) at the site of translocation. RACK1 is the selective RACK for PKCβII. In addition to anchoring activated PKC, RACK1 anchors other signaling enzymes such as sarcoma Src tyrosine kinase, integrin and phosphodiesterase 4D5. The ability of RACK1 to interact simultaneously with different signaling molecules, allows it to integrate inputs from distinct signaling pathways. In mammals, it regulates several processes that involve contact with extracellular matrix such as spreading, the establishment of focal adhesions and cell-cell contacts. Accumulating evidence attributes to it the role of regulating several major nervous system pathways. In addition, RACK1 is a ribosomal protein and a component small subunit and interacts ribosomal RNA. It recruits activated PKC to the ribosome, which in turn, phosphorylates initiation factor 6, leading to ribosome assembly.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interaction of Src and RACK1 is enhanced by activation of protein kinase C and tyrosine phosphorylation of RACK1
Chang BY, et al.
The Journal of Biological Chemistry, 276(23), 20346-20356 (2001)
Release of eIF6 (p27 BBP) from the 60S subunit allows 80S ribosome assembly
Ceci M, et al.
Nature, 426(6966), 579-579 (2003)
RACK1 has the nerve to act: structure meets function in the nervous system
Sklan EH, et al.
Progress in Neurobiology, 78(2), 117-134 (2006)
RACK1, A multifaceted scaffolding protein: Structure and function
Adams D R, et al.
Cell communication and signaling : CCS, 9(1), 22-22 (2011)
Xi-Xi Cao et al.
Breast cancer research and treatment, 126(3), 555-563 (2010-05-26)
We aimed to gain a mechanistic understanding of the role of RACK1 in breast carcinoma migration/metastasis. Migration assays were conducted in breast carcinoma cell lines. siRNA targeting RACK1 as well as the Rho kinase inhibitor were also applied. Immunoprecipitation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service