Skip to Content
Merck
All Photos(1)

Key Documents

PZ0221

Sigma-Aldrich

PF-06439015 methanesulfonate salt

≥98% (HPLC)

Synonym(s):

2R-(5-{6-Amino-5-[1R-(5-fluoro-2-[1,2,3]triazol-2-yl-phenyl)-ethoxy]-pyridin-3-yl}-4-methyl-thiazol-2-yl)-propane-1,2-diol methanesulfonate, PF-06439015 mesylate, PF-06439015 methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23FN6O3S · CH3SO3H
CAS Number:
Molecular Weight:
566.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

OC[C@](O)(C)C(S1)=NC(C)=C1C2=CN=C(N)C(O[C@H](C)C3=C(N4N=CC=N4)C=CC(F)=C3)=C2.CS(O)(=O)=O

InChI

1S/C22H23FN6O3S.CH4O3S/c1-12-19(33-21(28-12)22(3,31)11-30)14-8-18(20(24)25-10-14)32-13(2)16-9-15(23)4-5-17(16)29-26-6-7-27-29;1-5(2,3)4/h4-10,13,30-31H,11H2,1-3H3,(H2,24,25);1H3,(H,2,3,4)/t13-,22-;/m1./s1

InChI key

CVPQOLHPYNCJHK-RSUIGRPMSA-N

Biochem/physiol Actions

PF-06439015 is a potent and selective inihibitor that overcomes clinical ALK (receptor tyrosine kinase anaplastic lymphoma kinase) mutations resistant to Crizotinib.
PF-06439015 is a potent and selective inihibitor that overcomes clinical ALK (receptor tyrosine kinase anaplastic lymphoma kinase) mutations resistant to Crizotinib. PF-06439015 is potent across a broad panel of ALK mutant cell lines with an IC50 of 6.6 nM for tumor growth inhibition in a crizotinib-resistant cell line (H3122-L1196M).

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service