Skip to Content
Merck
All Photos(1)

Key Documents

PZ0016

Sigma-Aldrich

Dofetilide

≥98% (HPLC), powder, hERG channel blocker

Synonym(s):

N-[4-[2-[Methyl[2-[4-[(methylsulfonyl)amino]phenoxy]ethyl]amino]ethyl]phenyl]methanesulfonamide, UK-68798

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27N3O5S2
CAS Number:
Molecular Weight:
441.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Dofetilide, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

CN(CCOc1ccc(NS(C)(=O)=O)cc1)CCc2ccc(NS(C)(=O)=O)cc2

InChI

1S/C19H27N3O5S2/c1-22(13-12-16-4-6-17(7-5-16)20-28(2,23)24)14-15-27-19-10-8-18(9-11-19)21-29(3,25)26/h4-11,20-21H,12-15H2,1-3H3

InChI key

IXTMWRCNAAVVAI-UHFFFAOYSA-N

Gene Information

human ... KCNH2(3757)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dofetilide is a Class III antiarrhythmic and hERG channel blocker. Dofetilide selectively blocks the rapid component of the delayed rectifier outward potassium current (IKr).

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanne Elming et al.
Expert opinion on pharmacotherapy, 4(6), 973-985 (2003-06-05)
Atrial fibrillation (AF) is the most common cardiac arrhythmia. Mortality, and especially morbidity caused by AF, are major and growing health problems in the western world. AF is strongly associated with arterial hypertension, congestive heart failure, valvular heart disease, ischaemic
R Al-Dashti et al.
The Canadian journal of cardiology, 17(1), 63-67 (2001-02-15)
Dofetilide is a new, pure class III antiarrhythmic agent that prolongs the refractory period and action potential duration without having beta-blocking or calcium channel-blocking properties, making it unique among established class III agents. Dofetilide is effective in converting atrial and
Oleg E Osadchii
Cardiovascular drugs and therapy, 26(6), 489-500 (2012-08-25)
Dofetilide is class III antiarrhythmic agent which prolongs cardiac action potential duration because of selective inhibition of I (Kr), the rapid component of the delayed rectifier K(+) current. Although clinical studies reported on proarrhythmic risk associated with dofetilide treatment, the
João F S Carvalho et al.
Journal of medicinal chemistry, 56(7), 2828-2840 (2013-03-12)
Drug-induced blockade of the human ether-a-go-go-related gene K(+) channel (hERG) represents one of the major antitarget concerns in pharmaceutical industry. SAR studies of this ion channel have shed light on the structural requirements for hERG interaction but most importantly may
Zhiyi Yu et al.
Toxicology and applied pharmacology, 274(1), 78-86 (2013-11-10)
Drugs that block the cardiac K(+) channel encoded by the human ether-à-go-go gene (hERG) have been associated with QT interval prolongation leading to proarrhythmia, and in some cases, sudden cardiac death. Because of special structural features of the hERG K(+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service