Skip to Content
Merck
All Photos(3)

Documents

PRS4401

Sigma-Aldrich

Anti-ZBP1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ZBP1 Antibody - Anti-ZBP1 antibody produced in rabbit, Zbp1 Antibody, Anti-DAI, Anti-DLM-1, Anti-Z-DNA binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 39-45 kDa

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBP1(81030)

General description

Z-DNA binding protein 1 (ZBP1), also known as interferon (IFN) γ-inducible gene, is a cytoplasmic DNA sensor. It belongs to the family of Z-DNA binding proteins. ZBP1 is majorly expressed in the small intestine and lymphatic tissues. The gene is located on human chromosome 20q13.3.

Immunogen

a 18 amino acid peptide from near the carboxy-terminus of human ZBP1.

Application

Anti-ZBP1 antibody produced in rabbit has been used for western blotting.

Biochem/physiol Actions

Z-DNA binding protein 1 (ZBP1) plays a major role in DNA mediated innate immune responses. It enhances nuclear factor k-light-chain-enhancer of activated B cells (NFκB) activation and proinflammatory cytokine production.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4401.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZBP1: innate sensor regulating cell death and inflammation
Kuriakose T and Kanneganti TD
Trends in Immunology, 39(2), 123-134 (2018)
Functional roles of pattern recognition receptors that recognize virus nucleic acids in human adipose-derived mesenchymal stem cells
Yu L, et al.
BioMed Research International, 2016 (2016)
Molecular Lesions of Insulator CTCF and Its Paralogue CTCFL (BORIS) in Cancer: An Analysis from Published Genomic Studies
Voutsadakis I
High Throughput, 7(4), 30-30 (2018)
Lili Yu et al.
BioMed research international, 2016, 9872138-9872138 (2017-01-21)
Human adipose-derived mesenchymal stem cells (hAD-MSCs) are mesenchymal stem cells with the capability to modulate immune responses. Evidence showing that hAD-MSCs could mediate innate immune responses through pattern recognition receptors (PRRs) is increasing. However, the roles of PRRs in regulating
Zhiqiang Zhang et al.
Nature immunology, 14(2), 172-178 (2012-12-12)
DDX41 is a sensor of intracellular double-stranded DNA (dsDNA) in myeloid dendritic cells (mDCs) that triggers a type I interferon response via the signaling adaptor STING. We identified the E3 ligase TRIM21 as a DDX41-interacting protein and found that knockdown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service