Skip to Content
Merck
All Photos(1)

Key Documents

P9172

Sigma-Aldrich

Pyronin Y

for NA electrophoresis

Synonym(s):

Pyronin Y, Pyronin G, Pyronin J

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2O
CAS Number:
Molecular Weight:
302.80
Colour Index Number:
45005
Beilstein:
3795789
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

form

powder

composition

Dye content, ≥45%

technique(s)

nucleic acid detection: suitable

mp

250-260 °C (lit.)

solubility

water: 10 mg/mL, dark red

suitability

suitable for nucleic acid staining

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2C=C3C=C\C(C=C3Oc2c1)=[N+](/C)C

InChI

1S/C17H19N2O.ClH/c1-18(2)14-7-5-12-9-13-6-8-15(19(3)4)11-17(13)20-16(12)10-14;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

INCIMLINXXICKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Pyronin Y has been used:
  • in the purification of quiescent and cycling hematopoietic stem cells and staining of bone marrow cells (HSCs)
  • as a component of the polychlorinated biphenyls (PCBS) solution for staining mammary gland side population cells (MG-SPs)
  • staining of CD34+ cells

Biochem/physiol Actions

Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

Preparation Note

This product is soluble in water 1 mg/ml, yielding a clear dark red solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senescence and apoptosis block hematopoietic activation of quiescent hematopoietic stem cells with short telomeres
Wang J, et al.
Blood, 124(22), 3237-3240 (2014)
Photosensitizing effects of the tricyclic heteroaromatic cationic dyes pyronin Y and toluidine blue O (tolonium chloride
Darzynkiewicz Z and Carter SP
Cancer research, 48(5) (1988)
A gel electrophoresis method for determining the relative binding constants of biologically active, intercalating fluorescent stains.
S M Hassur et al.
Analytical biochemistry, 95(2), 329-339 (1979-06-01)
Green, F.J., ed.
The Sigma-Aldrich Handbook of Stains, Dyes and Indicators,, 601-602 (1990)
Staining of RNA after polyacrylamide gel electrophoresis.
K Marcinka
Methods in cell biology, 10, 49-68 (1975-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service