Skip to Content
Merck
All Photos(1)

Key Documents

P8180

Sigma-Aldrich

DL-α-Phosphatidylcholine, distearoyl

≥98%

Synonym(s):

1,2-Distearoyl-rac-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H88NO8P
CAS Number:
Molecular Weight:
790.15
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C44H89NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3,(H,48,49)

InChI key

OIBGDBRKAPVPMZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

İpek Eroğlu et al.
Journal of liposome research, 30(3), 263-273 (2019-06-13)
Acne vulgaris is one of the most common chronic diseases worldwide with the high prevalence ratio of about 80-85% in patients who are in puberty period. For the treatment options, many conventional dosage forms are available; however, existing limitations of
Saumyabrata Mazumder et al.
PLoS neglected tropical diseases, 5(12), e1429-e1429 (2011-12-30)
Vaccines that activate strong specific Th1-predominant immune responses are critically needed for many intracellular pathogens, including Leishmania. The requirement for sustained and efficient vaccination against leishmaniasis is to formulate the best combination of immunopotentiating adjuvant with the stable antigen (Ag)
Yoshiyuki Hattori et al.
Chemical & pharmaceutical bulletin, 59(11), 1386-1392 (2011-11-02)
Histone deacetylase inhibitor (HDACI), suberoylanilide hydroxamic acid (SAHA), approved by the Food and Drug Administration (FDA) for the treatment of cutaneous T cell lymphoma, is a promising new treatment strategy for various cancers. In this study, we hypothesized that a
Mette Marie Bruun Nielsen et al.
Langmuir : the ACS journal of surfaces and colloids, 29(5), 1525-1532 (2013-01-04)
Imaging ellipsometry (IE) has been applied to generate laterally resolved thickness maps of spin-coated membranes in both the dry and fully hydrated state. Spin-coating offers a convenient preparation method for stacked supported membranes, and in-depth thickness maps for such films
M Delwar Hussain et al.
International journal of pharmaceutics, 422(1-2), 290-294 (2011-11-22)
Solid dispersions of ibuprofen with various phospholipids were prepared, and the effect of phospholipids on the in vitro dissolution and in vivo gastrointestinal toxicity of ibuprofen was evaluated. Most phospholipids improved the dissolution of ibuprofen; dimyristoylphosphatidyl-glycerol (DMPG) had the greatest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service