Skip to Content
Merck
All Photos(3)

Key Documents

P8125

Sigma-Aldrich

Peroxidase from horseradish

Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)

Synonym(s):

Donor:hydrogen-peroxide oxidoreductase, Horseradish peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
Pricing and availability is not currently available.

type

Type I

form

essentially salt-free, lyophilized powder

specific activity

≥50 units/mg solid (using pyrogallol)

mol wt

~44 kDa

solubility

0.1 M phosphate buffer: soluble (pH 6.0)
H2O: soluble

absorbance ratio

RZ ≥1.0

storage temp.

2-8°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Horseradish peroxidase is isolated from horseradish roots (Amoracia rusticana) and belongs to the ferroprotoporphyrin group of peroxidases. HRP is a single chain polypeptide containing four disulfide bridges. It is a glycoprotein containing 18% carbohydrate. The carbohydrate composition consists of galactose, arabinose, xylose, fucose, mannose, mannosamine, and galactosamine depending upon the specific isozyme. Its molecular weight (~44 kDa) includes the polypeptide chain (33,890 Daltons), hemin plus Ca2+ (~700 Daltons), and carbohydrate (~9,400 Daltons). At least seven isozymes of HRP exist. The isoelectric point for horseradish peroxidase isozymes ranges from 3.0 - 9.0.

Application

Peroxidase from horseradish has been used:
  • in fecal starch analysis[1]
  • to measure plasma glucose levels by Trinder method[2]
  • to incubate feruloylated arabinoxylan fractions and also to evaluate oxidative coupling[3]
  • in the detection of glycoproteins[4]

Biochem/physiol Actions

HRP readily combines with hydrogen peroxide (H2O2) and the resultant [HRP-H2O2] complex can oxidize a wide variety of hydrogen donors. The optimal pH is 6.0-6.5 and the enzyme is most stable in the pH range of 5.0-9.0. HRP can be conjugated to antibodies by several different methods including glutaraldehyde, periodate oxidation, through disulfide bonds, and also via amino and thiol directed cross-linkers. It is smaller and more stable than the enzyme labels β-galactosidase and alkaline phosphatase.Hence, it is the most desired label. Also, its glycosylation leads to lower non-specific binding.[5] It is also used for the determination of glucose[6] and peroxides[7] in solution. Sodium azide, cyanide, L-cystine, dichromate, ethylenethiourea, hydroxylamine, sulfide, vanadate, p-aminobenzoic acid, and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+, Pb2+ ions are known to inhibit the enzyme activity.[8]
When incubated with a substrate, horseradish peroxidase produces a coloured, fluorimetric, or luminescent derivative of the labeled molecule, allowing quantification.

Unit Definition

One pyrogallol unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Analysis Note

The RZ (Reinheitszahl) is the absorbance ratio A403/A275 determined at 0.5-1.0 mg/ml in deionized water. It is a measure of hemin content, not enzymatic activity. Even preparations with high RZ may have low enzymatic activity.

Other Notes

View more information on peroxidase at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Allen et al.
Diabetes, 55(9), 2523-2533 (2006-08-29)
Halofenate has been shown previously to lower triglycerides in dyslipidemic subjects. In addition, significant decreases in fasting plasma glucose were observed but only in type 2 diabetic patients. We hypothesized that halofenate might be an insulin sensitizer, and we present
Studies on the oxidative cross-linking of feruloylated arabinoxylans from wheat flour and wheat bran
Schooneveld-Bergmans M E F, et al.
Carbohydrate Polymers, 38(4), 309-317 (1999)
Mirian M da Silva et al.
American journal of physiology. Heart and circulatory physiology, 285(1), H154-H162 (2003-03-08)
Ischemic preconditioning, or the protective effect of short ischemic episodes on a longer, potentially injurious, ischemic period, is prevented by antagonists of mitochondrial ATP-sensitive K+ channels (mitoKATP) and involves changes in mitochondrial energy metabolism and reactive oxygen release after ischemia.
Eva-Maria Herrlinger et al.
Chembiochem : a European journal of chemical biology, 21(16), 2329-2347 (2020-04-01)
Lysine-specific demethylase 1 (LSD1) has evolved as a promising therapeutic target for cancer treatment, especially in acute myeloid leukaemia (AML). To approach the challenge of site-specific LSD1 inhibition, we developed an enzyme-prodrug system with the bacterial nitroreductase NfsB (NTR) that
Acarbose enhances human colonic butyrate production
Weaver G A, et al.
The Journal of Nutrition, 127(5), 717-723 (1997)

Questions

  1. Hello! I am hoping to purchase this horseradish peroxidase to activate catharanthine in a reaction assay. How does the specific activity (pyrogallol units) translate to other molecules? And how should I calculate the amount of HRP to add to the reaction?

    1 answer
    1. Unfortunately, this product has not been tested for use in catharanthine activation. The end user would have to determine the appropriate amount experimentally. Please see the link below to review the product datasheet for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/259/166/p8125dat-ms.pdf

      Please see the link below to review a publication that may be helpful:
      https://www.sciencedirect.com/science/article/pii/S0141022997000677

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service