Skip to Content
Merck
All Photos(2)

Key Documents

P7876

Sigma-Aldrich

DL-Norephedrine hydrochloride

≥99%

Synonym(s):

(±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
CAS Number:
Molecular Weight:
187.67
Beilstein:
3914299
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

form

powder

mp

194-196 °C (lit.)

SMILES string

Cl.C[C@@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9-;/m1./s1

InChI key

DYWNLSQWJMTVGJ-PRCZDLBKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mixed-acting sympathomimetic amine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Sullivan et al.
European urology, 36 Suppl 1, 89-95 (1999-07-07)
Many patients with incontinence do not need surgery - for these patients symptoms can often be considerably improved by conservative measures, including drugs. Several different pharmacological actions are potentially useful depending on the underlying cause of the incontinence: a) Detrusor
Mona Karout et al.
Journal of neurochemistry, 139(5), 782-794 (2016-06-04)
The natural neurosteroid allopregnanolone exerts beneficial effects in animal models of neurodegenerative diseases, nervous system injury and peripheral neuropathies. It not only has anti-apoptotic activity, but also promotes proliferation of progenitor cells. With respect to using it as a therapeutic
R Dowse et al.
Journal of pharmaceutical sciences, 72(9), 1018-1020 (1983-09-01)
A high-performance liquid chromatographic analysis of phenylpropanolamine in human serum and urine without prior derivatization is presented. Using direct UV detection the method is sufficiently sensitive to detect 25 ng of drug/ml of serum or urine; the coefficients of variation
Tung Si Ho et al.
Journal of chromatographic science, 44(6), 308-316 (2006-08-04)
Different organic borates, phosphates, sulphates, and carboxylic acids are evaluated as extraction carriers in three-phase liquid-phase microextraction (LPME). Hydrophilic basic drugs form ion-pairs with the carriers and are extracted as ion-pair complexes into an organic liquid membrane of n-octanol or
Abraham G Sarraf et al.
International journal of pharmaceutics, 481(1-2), 9-17 (2014-12-30)
The objective of this study was to tailor a drug release profile through the adjustment of some key processing parameters involved in melt-extrusion: die temperature, shear rate, die length and drug particle size. Two experimental designs were selected, namely a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service