Skip to Content
Merck
All Photos(1)

Key Documents

P7181

Sigma-Aldrich

Polyethylene glycol solution

Hybri-Max, 50 % (w/v), average mol wt 1,450, 0.2 μm filtered, BioReagent, suitable for hybridoma

Synonym(s):

PEG Solution, Polyethylene Glycol

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

grade

Hybri-Max

Quality Level

sterility

0.2 μm filtered

product line

BioReagent

form

solution

mol wt

average mol wt 1,450

concentration

50 % (w/v)

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

Polyethylene glycol (PEG) is a polymer of ethylene oxide and water, obtained upon condensation.[1]

Application

Polyethylene glycol solution has been used in the hybridization of the following:
  • irradiated camel fibroblast cells with A23 hamster cells[2]
  • mouse splenocytes with Sp2/0-Ag 14 mouse myeloma cells[3]
  • rabbit splenocytes and cell line 240E-1[4]

Recommended for use in a normal fusion protocol requiring 50% PEG.

PEG is used as a fusogen to obtain hybridomas for monoclonal antibody production. Induces cell hybridization.

Biochem/physiol Actions

Polyethylene glycol (PEG) is used in detergents, emulsifying agents, soaps, metal costing process, pharmaceutical preparations and cosmetics.[1] PEG is commonly used as a lubricant. It′s application is also observed in chromatographic stationary phases.[5] PEG prevents crystal formation in emulsions.[6]

Packaging

Packaged in sealed ampules under nitrogen.

Other Notes

Contains 50% (w/v) polyethylene glycol (Av. Mol. Wt. 1450) in DPBS without calcium.

Reconstitution

Solution is ready-to-use. If a less concentrated solution is desired, dilute with sterile DPBS without calcium (D5773). Some precipitate may appear after being exposed to cooler, but should disappear as the solution warms. Solution may be frozen if desired but should first be aliquotted to avoid repeated freeze/thaw cycles.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a large panel of rabbit monoclonal antibodies against HIV-1 gp120 and isolation of novel neutralizing antibodies against the V3 loop
Qin Y, et al.
PLoS ONE, 10(6), e0128823-e0128823 (2015)
Development of Monoclonal Antibodies against CMP-N-Acetylneuraminate-beta-galactosamide-alpha-2, 3-sialyltransferase 1 (ST3Gal-I) Recombinant Protein Expressed in E. coli
Gupta AK, et al.
Biochemistry Research International, 2015(1), 1982-1982 (2015)
Construction of two whole genome radiation hybrid panels for dromedary (Camelus dromedarius): 5000 RAD and 15000 RAD
Perelman PL, et al.
Scientific reports, 8(1), 1982-1982 (2018)
Retinol encapsulated into amorphous Ca2+ polyphosphate nanospheres acts synergistically in MC3T3-E1 cells
Muller WEG, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 93(2), 214-223 (2015)
Thermal/oxidative degradation and stabilization of polyethylene glycol
Han S, et al.
Polymer, 38(2), 317-323 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service