Skip to Content
Merck
All Photos(1)

Key Documents

P7002

Sigma-Aldrich

Phospho(enol)pyruvic acid trisodium salt hydrate

≥97% (enzymatic), powder

Synonym(s):

2-(Phosphonooxy)-2-propenoic acid trisodium salt, PEP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H2O6PNa3 · x H2O
CAS Number:
Molecular Weight:
233.99 (anhydrous basis)
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)

Assay

≥97% (enzymatic)

form

powder

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

[Na].OC(=O)C(=C)OP(O)(O)=O

InChI

1S/C3H5O6P.Na.H/c1-2(3(4)5)9-10(6,7)8;;/h1H2,(H,4,5)(H2,6,7,8);;

InChI key

KZPUEZVJSLLABH-UHFFFAOYSA-N

Application

Phospho(enol)pyruvic acid trisodium salt hydrate has been used in:
  • several enzyme activity assays, such as PRPP synthetase (PRPPS) assay, ribokinase (RK) assay, adenine phosphoribosyltransferase (APRT) assay
  • in the reaction mixture for enzyme kinetic assay to determine platelet-derived growth factor receptor α (PDGFRA) kinetic parameters
  • as a component in buffer B to react with adenosine diphosphate (ADP) to form adenosine triphosphate (ATP) for energy charge measurement

Biochem/physiol Actions

Phospho(enol)pyruvic acid (PEP) is involved in glycolysis and gluconeogenesis. In glycolysis, PEP is metabolized by pyruvate kinase to yield pyruvate. In plants, PEP is involved in the formation of aromatic amino acids as well as in the carbon fixation pathway.
Phosphoenolpyruvic acid plays a major role in the initial step of glycolysis by acting as an inhibitor of hexokinase, phosphoglucose isomerase, phosphofructokinase and aldolase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deuterated nucleotides as chemical probes of RNA structure: a detailed protocol for the enzymatic synthesis of a complete set of nucleotides specifically deuterated at ribose carbons
Azad R, et al.
ScienceOpen Research (2015)
Kento Tokuyama et al.
Biotechnology and bioengineering, 115(6), 1542-1551 (2018-02-20)
Gene deletion strategies using flux balance analysis (FBA) have improved the growth-coupled production of various compounds. However, the productivities were often below the expectation because the cells failed to adapt to these genetic perturbations. Here, we demonstrate the productivity of
Structural and biochemical studies of the PDGFRA kinase domain
Liang L, et al.
Biochemical and Biophysical Research Communications, 477(4), 667-672 (2016)
Casimir Ledoux Sofeu et al.
PLoS neglected tropical diseases, 12(9), e0006597-e0006597 (2018-09-07)
Canine rabies is endemic in Cameroon, but human rabies exposures and cases are likely underreported because of inadequate surveillance. In 2014, the surveillance network in the West region of Cameroon was reinforced by introducing a new anti-rabies center, a framework
Energy metabolic state in hypothermically stored boar spermatozoa using a revised protocol for efficient ATP extraction
Nguyen QT, et al.
Biology Open, 5(11), 1743-1751 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service