Skip to Content
Merck
All Photos(1)

Key Documents

P5936

Sigma-Aldrich

Pectolyase from Aspergillus japonicus

greener alternative

suitable for plant cell culture, lyophilized powder

Synonym(s):

PEL1

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
10171502
NACRES:
NA.72
Pricing and availability is not currently available.

form

lyophilized powder

specific activity

≥0.3 unit/mg solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | plant: suitable

application(s)

agriculture

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been used as enzyme for alternative energy. For more information see the Enzymes for Alternative Energy Research.

Application

Pectolyase P5936 (Pel1) is a natural mixed pectolyase produced by the fungus Aspergillus japonicus used to digest components (endopolygalacturonate and pectin) of plant cell walls. Treatment of cell walls with pectolyase can be used to destablilize the cell walls for organelle isolation or to modify the elasticity of the cell walls.
Pectolyase from Aspergillus japonicas has been used to digest the siliques[1] and also in immunofluorescence experiments.[2][3]
Used in plant protoplast preparation to digest cell wall prior to organelle isolation.

Other Notes

Reported to contain two types of pectinase, endopolygalacturonase (EC3.2.1.15), endopectin lyase (EC4.2.2.10) and a maceration stimulating factor.

Unit Definition

One unit will liberate 1.0 μmole of galacturonic acid from polygalacturonic acid per min at pH 5.5 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AtKP1, a kinesin-like protein, mainly localizes to mitochondria in Arabidopsis thaliana
Ni C Z, et al.
Cell Research, 15(9), 725-725 (2005)
Detection of the quantity of kinesin and microgravity-sensitive kinesin genes in rat bone marrow stromal cells grown in a simulated microgravity environment
Ni C, et al.
Acta Astronautica, 68(11-12), 1722-1728 (2011)
Qingxin Zhao et al.
Plant physiology, 147(4), 1874-1885 (2008-06-20)
Mature plant cell walls lose their ability to expand and become unresponsive to expansin. This phenomenon is believed to be due to cross-linking of hemicellulose, pectin, or phenolic groups in the wall. By screening various hydrolytic enzymes, we found that
Nuclear fusions contribute to polyploidization of the gigantic nuclei in the chalazal endosperm of Arabidopsis
Baroux C, et al.
Planta, 220(1), 38-46 (2004)
Thorben Sprink et al.
International journal of molecular sciences, 22(17) (2021-09-11)
In the past, major findings in meiosis have been achieved, but questions towards the global understanding of meiosis remain concealed. In plants, one of these questions covers the need for two diverse meiotic active SPO11 proteins. In Arabidopsis and other

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service