Skip to Content
Merck
All Photos(2)

Key Documents

P5575

Sigma-Aldrich

Picloram

BioReagent, suitable for plant cell culture

Synonym(s):

4-Amino-3,5,6-trichloropyridine-2-carboxylic acid, 4-Amino-3,5,6-trichloropicolinic acid, Picloram

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3N2O2
CAS Number:
Molecular Weight:
241.46
Beilstein:
479075
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

form

powder

technique(s)

cell culture | plant: suitable

mp

200 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

Nc1c(Cl)c(Cl)nc(C(O)=O)c1Cl

InChI

1S/C6H3Cl3N2O2/c7-1-3(10)2(8)5(9)11-4(1)6(12)13/h(H2,10,11)(H,12,13)

InChI key

NQQVFXUMIDALNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Picloram has been used as a supplement in the growth media for culturing embryos.

Biochem/physiol Actions

Picloram (4-Amino-3,5,6-trichloropyridine-2-carboxylic acid) is a chlorinated systemic herbicide widely used for woody plant and broad-leaved weed control. Picloram induces direct somatic embryogenesis of Lilium longiflorum var. Ceb-dazzel.

Preparation Note

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient and reproducible Agrobacterium-mediated transformation method for hexaploid wheat (Triticum aestivum L.)
Hayta S, et al.
Plant methods, 15(1), 121-121 (2019)
D A Steinmacher et al.
Annals of botany, 108(8), 1463-1475 (2011-03-01)
Secondary somatic embryogenesis has been postulated to occur during induction of peach palm somatic embryogenesis. In the present study this morphogenetic pathway is described and a protocol for the establishment of cycling cultures using a temporary immersion system (TIS) is
Lin Tang et al.
Environmental science & technology, 42(4), 1207-1212 (2008-03-21)
Picloram, a widely used chlorinated herbicide, is quite persistent and mobile in soil and water with adverse health and environmental effects. It is essential to establish a rapid and sensitive method for accurate detection of trace picloram in agricultural samples.
Ali Ozcan et al.
Journal of hazardous materials, 153(1-2), 718-727 (2007-10-16)
The degradation of the picloram, a widely used herbicide, has been undertaken by the electrochemical advanced oxidation process, namely electro-Fenton in aqueous solution. This process generates catalytically hydroxyl radicals that are strong oxidizing reagents for the oxidation of organic substances.
Lijuan Chen et al.
Analytical biochemistry, 407(2), 172-179 (2010-08-17)
Picloram, a herbicide widely used for broadleaf weed control, is persistent and mobile in soil and water with adverse health and environmental effects. It is important to develop a sensitive method for accurate detection of trace picloram in the environment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service