Skip to Content
Merck
All Photos(1)

Key Documents

P5069

Sigma-Aldrich

Prostaglandin F tris salt

synthetic, suitable for cell culture

Synonym(s):

(5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt, PGF−Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5 · C4H11NO3
CAS Number:
Molecular Weight:
475.62
Beilstein:
4087514
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic
form:
powder
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

ethanol: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.[H][C@]1(C\C=C/CCCC(O)=O)[C@@H](O)C[C@@H](O)[C@]1([H])\C=C\[C@@H](O)CCCCC

InChI

1S/C20H34O5.C4H11NO3/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25;5-4(1-6,2-7)3-8/h4,7,12-13,15-19,21-23H,2-3,5-6,8-11,14H2,1H3,(H,24,25);6-8H,1-3,5H2/b7-4-,13-12+;/t15-,16+,17+,18-,19+;/m0./s1

InChI key

IYGXEHDCSOYNKY-RZHHZEQLSA-N

Gene Information

human ... PTGFR(5737)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds.
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. A positive feedback loop between endometrial PG F and luteal oxytocin is responsible for completion of luteolysis.
Luteolytic prostaglandin that induces parturition. Potent vasoconstrictor of pulmonary, coronary, and cerebral arteriole beds. Also stimulates contraction of smooth muscle, such as in bronchi, with possible involvement in asthma. This effect is at least partially mediated through tachykinin release.
Prostaglandin F2α is induced by uterine-produced oxytocin and acts on the corpus luteum to cause luteolysis and inhibit progesterone production.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Elmes et al.
Physiological reports, 3(4) (2015-04-17)
Advanced maternal age of first time pregnant mothers is associated with prolonged and dysfunctional labor and significant risk of emergency cesarean section. We investigated the influence of maternal age on myometrial contractility, expression of contractile associated proteins (CAPs), and global
J V Lopez et al.
The Veterinary record, 164(26), 807-809 (2009-06-30)
Twenty-four to 36 hours after farrowing, 192 sows were treated with a single intramuscular injection (2 ml per animal) of a prostaglandin analogue; 102 were treated with cloprostenol racemate and 97 with dinoprost tromethamine, and 90 were left untreated. There
M Sarkar et al.
Reproduction in domestic animals = Zuchthygiene, 45(3), 393-398 (2009-09-09)
An attempt was undertaken to investigate the efficacy of Ovsynch protocol for timed artificial insemination (TAI) with or without Norprolac (antiprolactin) treatment during non-breeding season (winter months) in yaks (n = 25). During non-breeding season, plasma prolactin profile has been
Augusta Zannoni et al.
Reproductive biology and endocrinology : RB&E, 5, 31-31 (2007-07-31)
The corpus luteum (CL) is a transient endocrine gland and prostaglandin F2-alpha is considered to be the principal luteolysin in pigs. In this species, the in vivo administration of prostaglandin F2-alpha induces apoptosis in large vessels as early as 6
Nicole A Rapicavoli et al.
BMC developmental biology, 10, 49-49 (2010-05-13)
Recent work has identified that many long mRNA-like noncoding RNAs (lncRNAs) are expressed in the developing nervous system. Despite their abundance, the function of these ncRNAs has remained largely unexplored. We have investigated the highly abundant lncRNA RNCR2 in regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service