Skip to Content
Merck
All Photos(1)

Key Documents

P4654

Sigma-Aldrich

Pentaethylene glycol monooctyl ether

Synonym(s):

C8E5, Octyl pentaethylene glycol ether, Octylpentaglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7(OCH2CH2)5OH
CAS Number:
Molecular Weight:
350.49
Beilstein:
1711907
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

Assay

≥98.0% (GC)

mol wt

350.49 g/mol

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCOCCOCCOCCOCCOCCO

InChI

1S/C18H38O6/c1-2-3-4-5-6-7-9-20-11-13-22-15-17-24-18-16-23-14-12-21-10-8-19/h19H,2-18H2,1H3

InChI key

MJELOWOAIAAUJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentaethylene glycol monooctyl ether has been used to study the size and morphology of aggregates. It has also been used in a study to investigate the sorption of monodisperse alcohol ethoxylates and their effects on the mobility of 2,4-D in isolated plant cuticles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria K Janowska et al.
Protein science : a publication of the Protein Society, 25(1), 286-294 (2015-09-04)
β-synuclein (βS) is a homologue of α-synuclein (αS), the major protein component of Lewy bodies in patients with Parkinson's disease. In contrast to αS, βS does not form fibrils, mitigates αS toxicity in vivo and inhibits αS fibril formation in
Study of the size and morphology of aggregates formed by pentaethylene glycol monooctyl ether (C"8EO"5) in n-heptane
Sandoval, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 17-23 (2012)
Sorption of Monodisperse Alcohol Ethoxylates and Their Effects on the Mobility of 2,4-D in Isolated Plant Cuticles
Riederer, M., et al.
Journal of Agricultural and Food Chemistry, 43, 1067-1075 (1995)
D R Bristow et al.
Journal of neurochemistry, 48(5), 1537-1540 (1987-05-01)
The detergent n-octylpentaoxyethylene is one in the series of tenside detergents developed for membrane solubilisation. We have used this detergent to solubilise benzodiazepine receptors from rat cerebellum. The soluble receptor has an affinity (KD) for [3H]flunitrazepam of 1.8 nM +/-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service