Skip to Content
Merck
All Photos(1)

Key Documents

P4235

Sigma-Aldrich

Anti-p53 antibody produced in sheep

fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53 kDa

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: 1-10 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TP53(7157)
mouse ... Trp53(22059)
rat ... Tp53(24842)

General description

p53 is a tumor suppressor gene expressed in a wide variety of tissues. It is a tetrameric nuclear DNA-binding phosphoprotein. The gene encoding it is localized on human chromosome 17p13.1 and mouse chromosome 11.

Immunogen

GST fusion protein of human p53 corresponding to amino acids 1-393.

Application

Monoclonal Anti-p53-Biotin antibody produced in mouse has been used for Western Blotting.

Biochem/physiol Actions

Tumor suppressor p53 has the capability to induce cell cycle arrest and has a role in DNA repair, senescence and apoptosis. It binds to Simian vacuolating virus 40 (SV40) T-antigen and human papilloma virus E6 protein. The p53 gene is mutated in various cancers, such as of the breast, ovarian, bladder, colon and lung.

Physical form

Solution in 0.01 M phosphate buffered saline, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell Cycle and Apoptosis
Bruna Pucci
Neoplasia, 2(4) (2000)
Cell Cycle and Apoptosis
Bruna Pucci
Neoplasia, 2(4) (2000)
ZnO nanorod-induced apoptosis in human alveolar adenocarcinoma cells via p53, survivin and bax/bcl-2 pathways: role of oxidative stress
Ahamad M
Nanomedicine: Nanotechnology, Biology, and Medicine, 7, 904-913 (2011)
Multiple myeloma involving central nervous system: high frequency of chromosome 17p13.1 (p53) deletions
Hong Chang
British Journal of Haematology, 127(3), 280?284-280?284 (2004)
Mutations in the p53 Tumor Suppressor Gene
Noa Rivlin
Genes & Cancer, 2(4), 466?474-466?474 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service