P4013
1,2-Dipalmitoyl-sn-glycero-3-phosphate sodium salt
≥99%
Synonym(s):
3-sn-Phosphatidic acid, 1,2-dipalmitoyl sodium salt
About This Item
Recommended Products
biological source
soybean
Assay
≥99%
form
powder
technique(s)
microbiological culture: suitable
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
SMILES string
[Na+].[Na+].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])([O-])=O)OC(=O)CCCCCCCCCCCCCCC
InChI
1S/C35H69O8P.2Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(36)41-31-33(32-42-44(38,39)40)43-35(37)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;;/h33H,3-32H2,1-2H3,(H2,38,39,40);;/q;2*+1/p-2/t33-;;/m1../s1
InChI key
NFRFUGBXJTXTMZ-QYKZUBHNSA-L
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service