Skip to Content
Merck
All Photos(1)

Key Documents

P3671

Sigma-Aldrich

Parathyroid Hormone Fragment 1-34 bovine

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C183H288N54O50S2
CAS Number:
Molecular Weight:
4108.71
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥70%

storage temp.

−20°C

InChI

1S/C174H278N54O49S2/c1-19-93(16)142(228-156(260)110(46-51-137(243)244)208-167(271)126(82-230)224-168(272)139(90(10)11)225-131(235)73-178)171(275)210-106(42-47-127(179)231)143(247)193-78-133(237)200-111(52-59-278-17)153(257)218-119(68-97-76-188-84-197-97)161(265)219-121(70-129(181)233)163(267)213-114(62-87(4)5)145(249)194-79-132(236)199-101(37-25-28-54-175)146(250)217-118(67-96-75-187-83-196-96)160(264)212-113(61-86(2)3)144(248)195-80-134(238)201-125(81-229)166(270)209-112(53-60-279-18)154(258)206-108(44-49-135(239)240)150(254)204-105(41-32-58-191-174(185)186)155(259)226-140(91(12)13)169(273)211-109(45-50-136(241)242)152(256)216-117(66-95-74-192-100-36-24-23-35-99(95)100)159(263)215-116(64-89(8)9)157(261)205-104(40-31-57-190-173(183)184)148(252)202-102(38-26-29-55-176)147(251)203-103(39-27-30-56-177)149(253)214-115(63-88(6)7)158(262)207-107(43-48-128(180)232)151(255)221-123(72-138(245)246)165(269)227-141(92(14)15)170(274)222-120(69-98-77-189-85-198-98)162(266)220-122(71-130(182)234)164(268)223-124(172(276)277)65-94-33-21-20-22-34-94/h20-24,33-36,74-77,83-93,101-126,139-142,192,229-230H,19,25-32,37-73,78-82,175-178H2,1-18H3,(H2,179,231)(H2,180,232)(H2,181,233)(H2,182,234)(H,187,196)(H,188,197)(H,189,198)(H,193,247)(H,194,249)(H,195,248)(H,199,236)(H,200,237)(H,201,238)(H,202,252)(H,203,251)(H,204,254)(H,205,261)(H,206,258)(H,207,262)(H,208,271)(H,209,270)(H,210,275)(H,211,273)(H,212,264)(H,213,267)(H,214,253)(H,215,263)(H,216,256)(H,217,250)(H,218,257)(H,219,265)(H,220,266)(H,221,255)(H,222,274)(H,223,268)(H,224,272)(H,225,235)(H,226,259)(H,227,269)(H,228,260)(H,239,240)(H,241,242)(H,243,244)(H,245,246)(H,276,277)(H4,183,184,190)(H4,185,186,191)/t93-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,139-,140-,141-,142-/m0/s1

InChI key

BHCZZGBILISWFT-KANWXXSKSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Val-Ser-Glu-Ile-Gln-Phe-Met-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe

General description

Parathyroid hormone (PTH) is a parathyroid-secreted polypeptide hormone that increases the level of calcium in blood by enhancing calcium mobilization from bone. It also increases the calcium: phosphate ratio in the kidney and promotes the absorption of calcium by the intestines. PTH acts as an anabolic agent that improves osteoblastic bone development.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jürgen Weisser et al.
Experimental cell research, 279(1), 1-13 (2002-09-06)
Differentiation and growth of chondrocytes in fetal growth plates of vertebrate long bones and ribs appear to occur in a gradual, continuous manner between the resting zone through the proliferation zone, maturation zone, and upper and lower hypertrophic zones, with
Renal control of calcium, phosphate, and magnesium homeostasis.
Blaine J
Clinical journal of the American Society of Nephrology : CJASN (2015)
J Green et al.
The American journal of physiology, 263(6 Pt 1), E1070-E1076 (1992-12-01)
Cytosolic free calcium ([Ca2+]i) is an important regulator of bone cell physiology. We studied the interaction of vitamin D metabolites on the hormonal-activated Ca message system in the osteoblastic cell line UMR-106. The acute rise in [Ca2+]i induced by different
G Klaus et al.
Endocrinology, 135(4), 1307-1315 (1994-10-01)
We investigated possible interaction of 1,25-dihydroxyvitamin D3 [1,25-(OH)2D3] and PTH on: 1) proliferation (monolayer culture) and colony formation (agarose stabilized suspension cultures); 2) expression of 1,25-(OH)2D3 receptor (VDR); and 3) cAMP response to PTH, using primary cultures of chondrocytes from
S Tetradis et al.
Endocrinology, 138(9), 3594-3600 (1997-09-01)
PTH increased PG synthase-2 transcription in osteoblastic MC3T3-E1 cells at 30 min, as assessed by nuclear run-on assays. To determine the signaling pathways used by PTH, the activity of a construct containing the PG synthase-2 gene between nucleotides -963 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service