Skip to Content
Merck
All Photos(5)

Key Documents

P3644

Sigma-Aldrich

L-α-Phosphatidylcholine

from soybean, Type IV-S, ≥30% (enzymatic)

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5209585
EC Number:
UNSPSC Code:
51321705
NACRES:
NA.25

biological source

soybean

type

Type IV-S

form

solid

concentration

≥30% (enzymatic)

solubility

chloroform: soluble 100 mg/mL, clear to slightly hazy, yellow to orange

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-α-Phosphatidylcholine is suitable for use:
  • as a component of human erythroid massive amplification culture (HEMAdef) medium for clinical expansion of human erythroblasts
  • in the chloride efflux assay to examine the chloride permeability of lipid vesicles conferred by soluble CLIC1
  • in an assay to measure solubilized wax synthase activity
  • as a component of buffer A used for the resuspension of the cholate-solubilized catalytic unit of adenylate cyclase
  • as a cryoprotectant for freezing ruminant sperm
  • as a substrate for the estimation of phospholipase activity
  • to make liposomes to assess their efficiency of recruitment of ARF (ADP ribosylation factor) and AP-1 (adaptor protein)
  • in the preparation of the isolated catalytic unit of soluble adenylate cyclase

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.
Phosphatidylcholine is the major membrane phospholipid in eukaryotic cells, forming the structural component of these membranes. It also serves as a reservoir for several lipid messengers and a source for several bioactive lipids such as, lysophosphatidylcholine, phosphatidic acid, diacylglycerol, lysophosphatidylcholine, platelet activating factor, and arachidonic acid.

Preparation Note

Purified from P 5638

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Andres Morales Letona et al.
Polymers, 11(2) (2019-04-10)
Here, we report a facile and effective one-pot approach to prepare uniform amylose-based polymeric microparticles (PMPs) through enzymatic synthesis of short-chain amylose (SCA) followed by spontaneous self-assembly of the SCA in the presence of lecithin. The effect of lecithin on
W Jiang et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(9), 4966-4971 (2001-04-26)
The stoichiometry of c subunits in the H(+)-transporting F(o) rotary motor of ATP synthase is uncertain, the most recent suggestions varying from 10 to 14. The stoichiometry will determine the number of H(+) transported per ATP synthesized and will directly
J W Winslow et al.
The Journal of biological chemistry, 261(17), 7626-7634 (1986-06-15)
ATP analogues were used to study the active site specificity of the catalytic unit (C) of solubilized and partially purified bovine brain caudate nucleus adenylate cyclase. Phenylenediamine ATP (PD-ATP), 8-azido ATP (8-N3ATP), chromium(III) 3'-beta-alanylarylazido ATP (CrATPa), and 2',3'-dialdehyde ATP (oATP)
Abouzar Najafi et al.
Animal reproduction science, 177, 35-41 (2016-12-25)
The aim of this study was to evaluate the effects of glycerol, ethylene glycol or DMSO in a soybean lecithin extender for freezing ram semen. In this study, 20 ejaculates were collected from four Ghezel rams and diluted with soybean
Glucanolytic enzyme production by Schizophyllum commune Fr. during mycoparasitism.
Chiu S C and Tzean S S.
Physiological & Molecular Plant Pathology, 46, 83-94 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service