Skip to Content
Merck
All Photos(1)

Key Documents

P2783

Sigma-Aldrich

Phosphotransacetylase from Bacillus stearothermophilus

lyophilized powder, ≥3,000 units/mg protein (biuret)

Synonym(s):

Acetyl-CoA:orthophosphate acetyltransferase, Phosphate acetyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥3,000 units/mg protein (biuret)

composition

Protein, ≥20%

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Phosphotransacetylase, from Bacillus stearothermophilus, is used to convert CoA to acetyl CoA. Phosphotransacetylase (pta) is used to study transport systems for acetate. It is used to study metabolic pathways in various bacterium.

Biochem/physiol Actions

Phosphotransacetylase converts CoA to acetyl CoA.

Unit Definition

One unit will convert 1.0 μmole of CoA to acetyl CoA per min at pH 7.5 at 30 °C using acetyl phosphate as substrate.

Physical form

Lyophilized powder containing potassium phosphate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutants of Bacillus stearothermophilus defective in the uptake and metabolism of acetate
Philip R. Mallinder and Anne Moir
Microbiology, 137, 779-785 (1991)
G D Sprott et al.
Applied and environmental microbiology, 59(4), 1092-1098 (1993-04-01)
Eleven strains of methanogenic bacteria were divided into two groups on the basis of the directionality (oxidative or reductive) of their citric acid pathways. These pathways were readily identified for most methanogens from the patterns of carbon atom labeling in
Ikuo Sato et al.
Bioscience, biotechnology, and biochemistry, 74(1), 75-81 (2010-01-09)
Malic enzyme (ME) was purified as an electrophoretically homogenous protein from Rhodopseudomonas palustris No. 7. The molecular weight of ME was estimated to be 650 kDa and that of its subunit, 86 kDa. ME activity was remarkably enhanced by di-
Guiqing Yang et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 26(2), 170-176 (2010-05-04)
Clostridium tyrobutyricum is suitable for simultaneous saccharification and fermentation of lignocellulosic. It can produce butyric acid, acetic acid as its main fermentation products from a wide variety of carbohydrates such as glucose, xylose, cellobiose and arabinose. In order to decrease
Ling Jiang et al.
Biotechnology and bioengineering, 108(1), 31-40 (2010-09-09)
Repeated fed-batch fermentation of glucose by Clostridium tyrobutyricum immobilized in a fibrous bed bioreactor (FBB) was successfully employed to produce butyric acid at a high final concentration as well as to adapt a butyric-acid-tolerant strain. At the end of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service