Skip to Content
Merck
All Photos(2)

Key Documents

P0453

Sigma-Aldrich

Piceatannol

powder

Synonym(s):

(E)-4-[2-(3,5Dihydroxyphenyl)ethenyl]1,2-benzenediol, 3,3′,4,5′-Stilbenetetrol, 3,3′,4,5′-Tetrahydroxy-trans-stilbene, Astringenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

color

light tan to yellow

mp

223.0-227.0 °C

solubility

H2O: 0.5 mg/mL
DMSO: 10 mg/mL
ethanol: 10 mg/mL

storage temp.

2-8°C

SMILES string

Oc1cc(O)cc(\C=C\c2ccc(O)c(O)c2)c1

InChI

1S/C14H12O4/c15-11-5-10(6-12(16)8-11)2-1-9-3-4-13(17)14(18)7-9/h1-8,15-18H/b2-1+

InChI key

CDRPUGZCRXZLFL-OWOJBTEDSA-N

Gene Information

human ... CDK2(1017)

Looking for similar products? Visit Product Comparison Guide

General description

Piceatannol is a natural tetrahydroxystilbene, that is extracted from grapes, red wine and Euphorbia lagascae seeds.

Application

Piceatannol has been used:
  • to study its effect on Ataxia-Telangiectasia Mutated (ATM) protein kinase
  • as an inhibitor of ZAP-70 (ζ-chain-associated protein kinase 70)
  • as an inhibitor of MRP1-mediated 2′,7′-bis-(carboxypropyl)-5(6)-carboxyfluorescein (BCPCF) transport in intact human erythrocytes and inside-out erythrocyte vesicles (IOVs)

Biochem/physiol Actions

Piceatannol is an analog of resveratrol that possess potential anti-oxidant, anti-cancer and anti-inflammatory properties. Picetannol has been shown to interfere with the cytokine signaling pathway, notably in the inhibition of the spleen tyrosine kinase (Syk) and PI3K. It also inhibits the activation of NFκB in human cultured cells, after treatment with several inflammatory agents, through inhibition of IκBα kinase and p65 phosphorylation.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Physical form

Photosensitive powder

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piceatannol, natural polyphenolic stilbene, inhibits adipogenesis via modulation of mitotic clonal expansion and insulin receptor-dependent insulin signaling in early phase of differentiation
Kwon JY, et al.
The Journal of Biological Chemistry, 287(14), 11566-11578 (2012)
José Pedro Lopes et al.
Frontiers in cellular and infection microbiology, 9, 289-289 (2019-08-29)
Cryptococcosis, caused by the basidiomycete Cryptococcus neoformans, is a life-threatening disease affecting approximately one million people per year worldwide. Infection can occur when C. neoformans cells are inhaled by immunocompromised people. In order to establish infection, the yeast must bypass
Nelly Noraz et al.
Development (Cambridge, England), 143(12), 2183-2193 (2016-04-29)
In the hematopoietic system, Syk family tyrosine kinases are essential components of immunoreceptor ITAM-based signaling. While there is increasing data indicating the involvement of immunoreceptors in neural functions, the contribution of Syk kinases remains obscure. Previously, we identified phosphorylated forms
Yiming Hao et al.
Nutrients, 11(7) (2019-07-07)
This study investigated the protective effect and the molecular mechanism of piceatannol on hydrogen peroxide (H2O2)-induced retinal pigment epithelium cell (ARPE-19) damage. Piceatannol treatment significantly inhibited H2O2-induced RPE cell death and reactive oxygen species (ROS) generation by 64.4% and 75.0%
Piceatannol inhibits TNF-induced NF-$\kappa$B activation and NF-$\kappa$B-mediated gene expression through suppression of I$\kappa$B$\alpha$ kinase and p65 phosphorylation
Ashikawa K, et al.
Journal of immunology (Baltimore, Md. : 1950), 169(11), 6490-6497 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service